    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO and CRO Solutions
          • Primary Antibodies ›
          • HIV1 p24 Antibodies
          Rockland
          HIV-1 p24 Monoclonal Antibody (7F4)
          View all (47) HIV1 p24 antibodies
          Tech Support
          Cite HIV-1 p24 Monoclonal Antibody (7F4)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          HIV-1 p24 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          HIV-1 p24 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          HIV-1 p24 Antibody (200-301-BR6) in WB
          Western Blot of Mouse anti-HIV-1 p24 antibody. Lane A: viral p24 protein at 0.5 µg/mL. Lane B: recombinant p24 protein at 1 µg/mL. Load: 20 ng. Primary antibody: HIV-1 p24 antibody overnight at 4°C. Secondary antibody: Mouse HRP secondary antibody. Block: 5% BLOTTO. Predicted/Observed size: 24, 41, 55 kDa, 24, 41, 55 kDa for HIV-1 p24. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          HIV-1 p24 Antibody in Western Blot (WB)
          HIV-1 p24 Antibody in ELISA (ELISA)
          Product Details
          200-301-BR6
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.2-0.5 µg/mL
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Virus
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          7F4
          Immunogen
          Anti- HIV-1 p24 antibody was produced in mice by repeated immunizations with a recombinant full-length HIV-1 p24 protein.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • Biotin 
          • HRP
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.2
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          HIV-1 Gag p24 is a capsid protein that constitutes the core of the AIDS virus HIV-1 and is produced by the digestion of its precursor Gag p55 by HIV-1 protease. The HIV-1 Gag p24 protein is indispensable to the reproduction of AIDS virus, and constitutes an essential element for the AIDS virus particle construction. HIV-1 Gag p24 is detectable from the early stage of AIDS virus infection, and is used as a marker for observation of the development in the patient's condition after treatment, as it indicated the amount of virus in the blood.
          Bioinformatics
          Protein Aliases: HIV p24; HIV1gp1
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline