    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
          • Order Status
          • 快速訂購
          • 登入
            登入
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • DYKDDDDK Tag Antibodies
          Rockland
          DYKDDDDK Monoclonal Antibody (29E4.G7), HRP
          View all (67) DYKDDDDK Tag antibodies
          Tech Support
          Cite DYKDDDDK Monoclonal Antibody (29E4.G7), HRP
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          DYKDDDDK Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DYKDDDDK Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          DYKDDDDK Antibody (200-303-383) in WB
          Western Blot of Mouse antibody for the detection of FLAG™ conjugated proteins Monoclonal Antibody Peroxidase Conjugated. Lane 1: FLAG™ Positive Control Lysates. Load: 10 µg per lane. Primary antibody: FLAG™ Monoclonal Antibody Peroxidase Conjugated at 1:1,000 for overnight at 4°C. Secondary antibody: none. Block: Blocking Buffer for Fluorescent Western Blotting for 30 min at RT. Predicted/Observed size: ~60 kDa, ~60 kDa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          DYKDDDDK Antibody in Western Blot (WB)
          Product Details
          200-303-383
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Immunohistochemistry (IHC)
          1:500-1:2,500
          -
          ELISA (ELISA)
          1:10,000-1:50,000
          -
          Immunoprecipitation (IP)
          1:10,000
          -
          Product Specifications
          Species Reactivity
          Tag
          Host/Isotype
          Mouse / IgG2a, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          29E4.G7
          Immunogen
          This antibody was produced in mice by repeated immunizations with a synthetic peptide corresponding to the FLAG™ epitope tag peptide DYKDDDDK (Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys) conjugated to KLH using maleimide. Residues of glycine and cysteine were added to the termini to facilitate coupling.
          Conjugate
          HRP HRP HRP
          • Unconjugated
          • Agarose
          • Biotin 
          • DyLight 405
          • DyLight 488
          • DyLight 549
          • DyLight 649
          • DyLight 680
          • DyLight 800
          • FITC
          • Request custom conjugation
          Form
          Lyophilized
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          0.02M potassium phosphate, pH 7.2, with 10mg/mL BSA, 0.15M NaCl
          Contains
          0.01% gentamicin sulfate
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Store vial at 4° C prior to restoration. For extended storage aliquot contents and freeze at -20° C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
          This antibody is directed against the FLAG™ epitope tag and is useful in determining its presence in over expressed proteins in various assays. The antibody recognizes the FLAG™ epitope tag (Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys) fused to either the amino- or carboxy- termini of targeted proteins in transfected or transformed cells.
          Target Information
          DYKDDDDK tag, also known as FLAG tag, is a short peptide sequence that is commonly used as a protein tag to facilitate protein purification, detection, and localization. The tag consists of eight amino acids: Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys. It is often fused to the N- or C-terminus of a protein of interest, allowing for easy identification and isolation of the protein using anti-FLAG antibodies. The DYKDDDDK tag is widely used in molecular biology and biochemistry research, and has been employed in various applications, such as protein-protein interaction studies, protein expression analysis, and protein localization studies. The FLAG tag structure has been optimized for compatibility with the proteins it is attached to, in that it is more hydrophilic than other common epitope tags and therefore less likely to denature or inactivate proteins to which it is appended. FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany.
          Bioinformatics
          Protein Aliases: DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline