    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO and CRO Solutions
          • Primary Antibodies ›
          • PTPRF Antibodies
          Rockland
          LAR/PTPRF Monoclonal Antibody (S165-38), HRP
          View all (23) PTPRF antibodies
          Tech Support
          Cite LAR/PTPRF Monoclonal Antibody (S165-38), HRP
          Additional Information:
          {{banner.description}}
          LAR/PTPRF Monoclonal Antibody (S165-38), HRP
          Product Details
          200-303-W57
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Immunohistochemistry (IHC)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          S165-38
          Immunogen
          Anti-LAR/PTPRF Antibody was produced in mice by repeated immunizations of a fusion peptide consisting of amino acids 1315-1607 of human LAR.
          3D Epitope / Immunogen
          Conjugate
          HRP HRP HRP
          • Unconjugated
          • Biotin 
          • FITC
          • PE 
          • PerCP 
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.2, with 50% glycerol
          Contains
          0.1% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          Possible cell adhesion receptor. It possesses an intrinsic protein tyrosine phosphatase activity (PTPase).The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate.
          Bioinformatics
          Protein Aliases: FLJ43335; FLJ45062; FLJ45567; LAR; LAR protein; LCA homolog; LCA-homolog; leukocyte antigen-related (LAR) PTP receptor; leukocyte antigen-related tyrosine phosphatase; Leukocyte common antigen related; protein tyrosine phosphatase receptor-type F; protein tyrosine phosphatase, receptor type, F polypeptide; PTPRF protein; receptor-linked protein-tyrosine phosphatase LAR; Receptor-type tyrosine-protein phosphatase F; unnamed protein product
          View more View less
          Gene Aliases: AA591035; BNAH2; LAR; LARFN5C; LARS; PTPRF; RPTP-LAR
          View more View less
          UniProt ID: (Human) P10586, (Rat) Q64604, (Mouse) A2A8L5
          View more View less
          Entrez Gene ID: (Human) 5792, (Rat) 360406, (Mouse) 19268
          View more View less
          Function(s)
          phosphoprotein phosphatase activity protein tyrosine phosphatase activity transmembrane receptor protein tyrosine phosphatase activity heparin binding hydrolase activity chondroitin sulfate proteoglycan binding macromolecular complex binding cell adhesion molecule binding insulin receptor binding receptor tyrosine kinase binding protein complex binding phosphate ion binding phosphatase activity protein phosphatase protein modifying enzyme
          Process(es)
          cell adhesion signal transduction transmembrane receptor protein tyrosine phosphatase signaling pathway nervous system development regulation of neuron projection development cell migration neuron projection regeneration peptidyl-tyrosine dephosphorylation regulation of axon regeneration synaptic membrane adhesion negative regulation of receptor binding negative regulation of cytokine-mediated signaling pathway protein dephosphorylation homophilic cell adhesion via plasma membrane adhesion molecules negative regulation of cell proliferation negative regulation of cell projection organization negative regulation of epidermal growth factor receptor signaling pathway positive regulation of apoptotic process positive regulation of neuron apoptotic process negative regulation of insulin receptor signaling pathway positive regulation of dendrite morphogenesis regulation of synapse structure or activity negative regulation of neurotrophin TRK receptor signaling pathway dephosphorylation
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline