    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Neuroligin 1 Antibodies
          Rockland
          Neuroligin 1 Monoclonal Antibody (S97A-31), PE
          View all (36) Neuroligin 1 antibodies
          Tech Support
          Cite Neuroligin 1 Monoclonal Antibody (S97A-31), PE
          Additional Information:
          {{banner.description}}
          Neuroligin 1 Monoclonal Antibody (S97A-31), PE
          Product Details
          200-308-W75
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500
          -
          Immunohistochemistry (IHC)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          S97A-31
          Immunogen
          Anti-Neuroligin 1 Antibody was produced by repeated immunization of mice with a fusion protein containing amino acids 718-843 of rat Neuroligin-1.
          3D Epitope / Immunogen
          Conjugate
          PE PE PE
          • Unconjugated
          • Biotin 
          • FITC
          • HRP
          • PerCP 
          • Request custom conjugation
          Excitation/Emission Max
          565/576 nm View spectra spectra
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.2, with 50% glycerol
          Contains
          0.1% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses.
          Bioinformatics
          Protein Aliases: MGC45115; Neuroligin I; Neuroligin-1; NLG 1; unnamed protein product
          View more View less
          Gene Aliases: 6330415N05Rik; BB179718; KIAA1070; mKIAA1070; NL1; NLG1; NLGN1
          View more View less
          UniProt ID: (Human) Q8N2Q7, (Mouse) Q99K10, (Rat) Q62765
          View more View less
          Entrez Gene ID: (Human) 22871, (Mouse) 192167, (Rat) 116647
          View more View less
          Function(s)
          beta-amyloid binding PDZ domain binding signaling receptor activity neurexin family protein binding identical protein binding cell adhesion molecule binding scaffold protein binding receptor activity protein binding protein dimerization activity carboxylic ester hydrolase activity cell adhesion molecule
          Process(es)
          protein targeting cell adhesion heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules neuron cell-cell adhesion nervous system development synapse assembly positive regulation of circadian sleep/wake cycle, wakefulness synaptic vesicle targeting calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules neuronal signal transduction neuron projection development positive regulation of synaptic transmission, GABAergic protein localization to synapse establishment of protein localization regulation of neuron differentiation rhythmic process cytoskeletal matrix organization at active zone neuron projection morphogenesis modulation of synaptic transmission positive regulation of filopodium assembly positive regulation of synapse assembly positive regulation of synaptic transmission, glutamatergic positive regulation of dendritic spine development negative regulation of dendritic spine morphogenesis cellular response to calcium ion terminal button organization synaptic vesicle clustering postsynaptic membrane assembly presynaptic membrane assembly AMPA glutamate receptor clustering NMDA glutamate receptor clustering neurexin clustering involved in presynaptic membrane assembly postsynaptic density protein 95 clustering receptor localization to synapse NMDA selective glutamate receptor signaling pathway AMPA selective glutamate receptor signaling pathway presynapse assembly neuron projection arborization positive regulation of ruffle assembly positive regulation of neuromuscular synaptic transmission positive regulation of synaptic vesicle endocytosis positive regulation of intracellular signal transduction excitatory synapse assembly positive regulation of synaptic vesicle exocytosis positive regulation of excitatory postsynaptic potential regulation of respiratory gaseous exchange by neurological system process synaptic vesicle transport synapse organization protein homooligomerization protein heterotetramerization inhibitory postsynaptic potential long-term synaptic potentiation positive regulation of protein localization to synapse regulation of N-methyl-D-aspartate selective glutamate receptor activity regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity positive regulation of synaptic vesicle clustering metabolic process neuronal ion channel clustering
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline