    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GST Antibodies
          Rockland
          GST Monoclonal Antibody (3D4), DyLight™ 488
          View all (103) GST antibodies
          Tech Support
          Cite GST Monoclonal Antibody (3D4), DyLight™ 488
          Additional Information:
          {{banner.description}}
          Product Details
          200-341-200
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:10,000-1:25,000
          -
          Immunocytochemistry (ICC/IF)
          1:500-1:2,500
          -
          ELISA (ELISA)
          >1:20,000
          -
          Product Specifications
          Species Reactivity
          Tag
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3D4
          Immunogen
          BALB/c mice were immunized with GST from full length Schistosoma japonicum protein. A hybridoma was produced by the fusion of BALB/c mouse splenocytes and myeloma cells using conventional hybridoma technology.
          Conjugate
          DyLight™ 488 DyLight™ 488 DyLight™ 488
          • Unconjugated
          • DyLight 405
          • DyLight 549
          • DyLight 649
          • DyLight 680
          • DyLight 800
          • Request custom conjugation
          Excitation/Emission Max
          492/519 nm View spectra spectra
          Form
          Lyophilized
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          0.02M potassium phosphate, pH 7.2, with 10mg/mL BSA, 0.15M NaCl
          Contains
          0.01% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Store vial at 4° C prior to restoration. For extended storage aliquot contents and freeze at -20° C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
          Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Mouse Serum and purified and partially purified Glutathione-S-Transferase (GST).
          Target Information
          GST (Glutathione S-transferase) is a 26kDa protein, present in eukaryotes and in prokaryotes, where they catalyze a variety of reactions. Glutathione Stransferase is used to create the GST gene fusion system. This GST-fusion protein can then be purified from cells via its high affinity for glutathione. Fusion Proteins offer an important biological assay for direct protein-to-protein interactions. The GST gene fusion system has been introduced into numerous expression vectors.
          Bioinformatics
          Protein Aliases: AT1G19550; F18O14.31; F18O14_31; Glutathione-S-Transferase; GST protein; GST Tag
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline