    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • GFP Antibodies
          Rockland
          GFP Monoclonal Antibody (9F9.F9), DyLight™ 680
          View all (134) GFP antibodies
          Tech Support
          Cite GFP Monoclonal Antibody (9F9.F9), DyLight™ 680
          Additional Information:
          {{banner.description}}
          Product Details
          200-344-215
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:10,000-1:25,000
          -
          Immunocytochemistry (ICC/IF)
          >1:5,000
          -
          ELISA (ELISA)
          >1:10,000
          -
          Product Specifications
          Species Reactivity
          Tag
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          9F9.F9
          Immunogen
          BALB/c mice were immunized with a Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246 aa) derived from the jellyfish Aequorea victoria.
          Conjugate
          DyLight™ 680 DyLight™ 680 DyLight™ 680
          • Unconjugated
          • AP
          • Biotin 
          • DyLight 405
          • DyLight 488
          • DyLight 549
          • DyLight 649
          • DyLight 800
          • FITC
          • HRP
          • Rhodamine
          • Request custom conjugation
          Excitation/Emission Max
          676/705 nm View spectra spectra
          Form
          Lyophilized
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          0.02M potassium phosphate, pH 7.2, with 10mg/mL BSA, 0.15M NaCl
          Contains
          0.01% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Store vial at 4° C prior to restoration. For extended storage aliquot contents and freeze at -20° C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
          Assay by Immunoelectrophoresis resulted in a single precipitin arc against anti-Mouse Serum. Reactivity is observed against recombinant Green Fluorescent Protein (000-001-215, recombinant GFP from Aequorea victoria) by Western blot. No reaction is seen against RFP.
          Target Information
          The jellyfish Aequorea victoria contains green fluorescent protein (GFP Tag) that emits light in the bioluminescence reaction of the animal. GFP has been used widely as a reporter protein for gene expression in eukaryotic and prokaryotic organisms, and as a protein tag in cell culture and in multicellular organisms. As a fusion tag, GFP can be used to localize proteins, to study their movement or to research the dynamics of the subcellular compartments where these proteins are targeted. GFP technology has revealed considerable new insights in the physiological activities of living cells. GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species independent and is suitable for a variety of applications. GFP has been used extensively as a fluorescent tag to monitor gene expressin and protein localization. Moreover, other applications for GFP include its use in assessing protein protein interactions in the yeast two hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.
          Bioinformatics
          Protein Aliases: GFP; GFP tag; GFP2; green fluorescence; green fluorescent; Turbo eGFP
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline