    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CEP55 Antibodies
          Proteintech
          CEP55 Polyclonal Antibody
          15 References
          View all (20) CEP55 antibodies
          Tech Support
          Cite CEP55 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (8)
          CEP55 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          CEP55 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 8
          CEP55 Antibody (23891-1-AP) in IHC (P)
          Immunohistochemistry of paraffin-embedded human testis tissue slide using 23891-1-AP ( CEP55 Antibody) at dilution of 1:50 (under 40x lens). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CEP55 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CEP55 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CEP55 Antibody in Western Blot (WB)
          CEP55 Antibody in Western Blot (WB)
          CEP55 Antibody in Western Blot (WB)
          CEP55 Antibody in Western Blot (WB)
          CEP55 Antibody in Western Blot (WB)
          CEP55 Antibody in Immunoprecipitation (IP)
          CEP55 Polyclonal Antibody
          Product Details
          23891-1-AP
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:2,000
          View 7 publications 7 publications
          Immunohistochemistry (IHC)
          -
          View 3 publications 3 publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1:20-1:200
          -
          Immunocytochemistry (ICC/IF)
          -
          View 3 publications 3 publications
          Flow Cytometry (Flow)
          Assay-dependent
          View 1 publication 1 publication
          Immunoprecipitation (IP)
          0.5-4.0 µg
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Published species
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          CEP55 Fusion Protein Ag20940 (1-355 aa encoded by BC008947)
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.42 mg/mL
          Amount
          63 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice (domestic); Dry ice (international)
          Product Specific Information
          Immunogen sequence: MSSRSTKDL IKSKWGSKPS NSKSETTLEK LKGEIAHLKT SVDEITSGKG KLTDKERHRL LEKIRVLEAE KEKNAYQLTE KDKEIQRLRD QLKARYSTTA LLEQLEETTR EGERREQVLK ALSEEKDVLK QQLSAATSRI AELESKTNTL RLSQTVAPNC FNSSINNIHE MEIQLKDALE KNQQWLVYDQ QREVYVKGLL AKIFELEKKT ETAAHSLPQQ TKKPESEGYL QEEKQKCYND LLASAKKDLE VERQTITQLS FELSEFRRKY EETQKEVHNL NQLLYSQRRA DVQHLEDDRH KTEKIQKLRE ENDIARGKLE EEKKRSEELL SQVQFLYTSL LKQQEEQTRV ALLEQQ (1-355 aa encoded by B C008947)
          Target Information
          Plays a role in mitotic exit and cytokinesis (PubMed:16198290, PubMed:17853893). Recruits PDCD6IP and TSG101 to midbody during cytokinesis. Required for successful completion of cytokinesis (PubMed:17853893). Not required for microtubule nucleation (PubMed:16198290). Plays a role in the development of the brain and kidney (PubMed:28264986). [UniProt]
          Bioinformatics
          Protein Aliases: cancer/testis antigen 111; centrosomal protein 55kDa; Centrosomal protein of 55 kDa; CEP55; unnamed protein product; Up-regulated in colon cancer 6
          View more View less
          Gene Aliases: 1200008O12Rik; 2700032M20Rik; C10orf3; CEP55; CT111; MARCH; RGD1305340; URCC6
          View more View less
          UniProt ID: (Human) Q53EZ4, (Mouse) Q8BT07, (Rat) Q4V7C8
          View more View less
          Entrez Gene ID: (Human) 55165, (Mouse) 74107, (Rat) 294074
          View more View less
          Function(s)
          protein binding identical protein binding
          Process(es)
          mitotic cytokinesis cell separation after cytokinesis cell cycle mitotic nuclear division establishment of protein localization cell division nucleus organization mitotic metaphase plate congression regulation of protein kinase B signaling midbody abscission cranial skeletal system development
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline