Immunogen sequence: KTFAPNTSL NSWDPICSAK EANIQRNLER SHWLTSYTHD FTGLGPMDPL ELDDYHEKMV AELTRKIGFD PEPQEKFHPV FKPPRPLEGR IARLIQNRRS LEAIVQQRPR SCPDCTPRVL CNFHTFVPSS KEMVALSDNI PAGVTHKNQD IEEKIIEEQS LLSTYELPSC YPTKDLTSIY DIKPFPKITD TKKTEDLYWR QQSLKTQPTP YCKPDHWIHY ENLKSPLRDQ YNMCPDPVSL SKPSVLQNKQ DTEAFTLEHF LSKPEEELFL NMENNEETRP VLGWIPRAGV TKPQTNLLEL KNSFSKTGAQ KRFHKSILED HKDLRDNEHS GMKHQFYGHN SYYFYN (246-590 aa encoded by BC043269)
C7orf31 is a protein coding gene.
Protein Aliases: sperm associated microtubule inner protein 4; Sperm-associated microtubule inner protein 4; uncharacterized protein C7orf31; unnamed protein product
Gene Aliases: C7orf31; SPMIP4
UniProt ID: (Human) Q8N865
Entrez Gene ID: (Human) 136895