    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • TRA-1-60 Antibodies
          Invitrogen
          TRA-1-60 Monoclonal Antibody (cl.A)
          6 Published Figures
          40 References
          View all (10) TRA-1-60 antibodies
          Tech Support
          Cite TRA-1-60 Monoclonal Antibody (cl.A)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          • Published Figures (6)
          TRA-1-60 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TRA-1-60 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 8
          TRA-1-60 Antibody (41-1000) in ICC/IF
          Indirect immunofluorescent analysis of SHEFF4 human ES cells using Zymed Ms anti-TRA-1-60 (Product # 41-1000). Image courtesy of Peter Andrews, Ph.D., University of Sheffield, Sheffield, UK. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TRA-1-60 Antibody in Immunocytochemistry (ICC/IF)
          TRA-1-60 Antibody in Western Blot (WB)
          TRA-1-60 Antibody in Immunocytochemistry (ICC/IF)
          TRA-1-60 Antibody in Immunocytochemistry (ICC/IF)
          TRA-1-60 Antibody in Immunocytochemistry (ICC/IF)
          TRA-1-60 Antibody in Immunocytochemistry (ICC/IF)
          TRA-1-60 Antibody in Immunocytochemistry (ICC/IF)
          TRA-1-60 Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          41-1000
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          View 2 publications 2 publications
          Immunohistochemistry (IHC)
          -
          View 2 publications 2 publications
          Immunohistochemistry (PFA fixed) (IHC (PFA))
          -
          View 1 publication 1 publication
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          View 28 publications 28 publications
          Flow Cytometry (Flow)
          -
          View 7 publications 7 publications
          Product Specifications
          Species Reactivity
          Human
          Published species
          Human, Mouse
          Host/Isotype
          Mouse / IgM
          Class
          Monoclonal
          Type
          Antibody
          Clone
          cl.A
          Immunogen
          2102Ep cl.2A6 human embryonal carcinoma cells
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography - MBP
          Storage buffer
          PBS, pH 7.4
          Contains
          0.1% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2533494
          Target Information
          TRA-1-60 is a cell surface antigen, expressed along with SSEA-3, SSEA-4 and TRA-1-81 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are lost during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.
          Bioinformatics
          Protein Aliases: TRA160; tra60
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline