    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DC-SIGN (CD209) Antibodies
          Invitrogen
          CD209a Monoclonal Antibody (MMD3), Alexa Fluor™ 488, eBioscience™
          1 Published Figure
          1 Reference
          View all (64) DC-SIGN (CD209) antibodies
          Tech Support
          Cite CD209a Monoclonal Antibody (MMD3), Alexa Fluor™ 488, eBioscience™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          • Published Figures (1)
          CD209a Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          CD209a Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          CD209a Antibody (53-2094-80) in IHC (F)
          Immunohistochemistry of frozen 5-week-old mouse thymus stained with 20 µg/mL of Mouse IgG2b K Isotype Control Alexa Fluor® 488 (left, green) or 20 µg/mL of Anti-Mouse CD209a (DC-SIGN) Alexa Fluor® 488 (right, green) and costained with 20 µg/mL of Anti-Mouse CD8a eFluor® 660 (red). Nuclei are stained with DAPI (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD209a Antibody in Immunohistochemistry (Frozen) (IHC (F))
          CD209a Antibody in Flow Cytometry (Flow)
          Product Details
          53-2094-80
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          20 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          Assay-Dependent
          -
          Flow Cytometry (Flow)
          -
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Mouse / IgG2c, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          MMD3
          Conjugate
          Alexa Fluor™ 488 Alexa Fluor™ 488 Alexa Fluor™ 488
          • eFluor 660
          • Request custom conjugation
          Excitation/Emission Max
          499/520 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          25 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.2
          Contains
          0.09% sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2574418
          Product Specific Information
          Description: This MMD3 monoclonal antibody recognizes mouse CD209a, which is also known as DC-SIGN. CD209a is a type II transmembrane C-type lectin expressed on a subset of dendritic cells, including some CD4+, CD8- and plasmacytoid pre-dendritic cells. Studies indicate that CD209a expression can vary according to the activation state of the host. Moreover, CD209a is down-regulated in spleen-derived dendritic cell cultures supplemented with GM-CSF. CD209a is involved in mediating the innate immune response by binding microbial carbohydrates.
          Cross-blocking studies suggest that MMD3 recognizes a different a different epitope from LWC06 and 5H10.
          Applications Reported: This MMD3 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, microscopy, immunohistochemical staining, and immunocytochemistry.
          Applications Tested: This MMD3 antibody has been tested by immunohistochemistry on frozen 5-week-old mouse thymus and can be used at less than or equal to 20 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
          Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.
          Filtration: 0.2 µm post-manufacturing filtered.
          Target Information
          This gene encodes a transmembrane receptor and is often referred to as DC-SIGN because of its expression on the surface of dendritic cells and macrophages. The encoded protein is involved in the innate immune system and recognizes numerous evolutionarily divergent pathogens ranging from parasites to viruses with a large impact on public health. The protein is organized into three distinct domains: an N-terminal transmembrane domain, a tandem-repeat neck domain and C-type lectin carbohydrate recognition domain. The extracellular region consisting of the C-type lectin and neck domains has a dual function as a pathogen recognition receptor and a cell adhesion receptor by binding carbohydrate ligands on the surface of microbes and endogenous cells. The neck region is important for homo-oligomerization which allows the receptor to bind multivalent ligands with high avidity. Variations in the number of 23 amino acid repeats in the neck domain of this protein are rare but have a significant impact on ligand binding ability. This gene is closely related in terms of both sequence and function to a neighboring gene (GeneID 10332; often referred to as L-SIGN). DC-SIGN and L-SIGN differ in their ligand-binding properties and distribution. Alternative splicing results in multiple variants.
          Bioinformatics
          Protein Aliases: CD209; CD209 antigen-like protein A; DC-SIGN; Dendritic cell-specific ICAM-3-grabbing non-integrin; MGC129965; MGC130443
          View more View less
          Gene Aliases: CD209; Cd209a; CDSIGN; CIRE; DC-SIGN; DC-SIGN1; Dcsign; SIGN-R1; SIGNR5
          View more View less
          UniProt ID: (Mouse) Q91ZX1
          View more View less
          Entrez Gene ID: (Mouse) 170786
          View more View less
          Function(s)
          virus receptor activity mannose binding carbohydrate binding metal ion binding calcium-dependent protein binding carbohydrate derivative binding membrane traffic protein
          Process(es)
          endocytosis regulation of gene expression regulation of blood coagulation regulation of T cell proliferation viral entry into host cell
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline