    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • Tartrate Resistant Acid Phosphatase Antibodies
          NeoBiotechnologies
          TRAcP (Tartrate-Resistant Acid Phosphatase) Recombinant Mouse Monoclonal Antibody (rACP5, 1070)
          View all (28) Tartrate Resistant Acid Phosphatase antibodies
          Tech Support
          Cite TRAcP (Tartrate-Resistant Acid Phosphatase) Recombinant Mouse Monoclonal Antibody (rACP5, 1070)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          TRAcP (Tartrate-Resistant Acid Phosphatase) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          TRAcP (Tartrate-Resistant Acid Phosphatase) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          TRAcP (Tartrate-Resistant Acid Phosphatase) Antibody (54-MSM3-P0) in IHC (P)
          Immunohistochemistry (Paraffin) analysis performed in formalin fixed, paraffin-embedded human spleen using TRAcP (Tartrate-Resistant Acid Phosphatase) Recombinant Monoclonal Antibody (Product # 54-MSM3-P1). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TRAcP (Tartrate-Resistant Acid Phosphatase) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TRAcP (Tartrate-Resistant Acid Phosphatase) Antibody in Peptide array (ARRAY)
          TRAcP (Tartrate-Resistant Acid Phosphatase) Antibody in SDS-PAGE (SDS-PAGE)
          TRAcP (Tartrate-Resistant Acid Phosphatase) Recombinant Mouse Monoclonal Antibody (rACP5, 1070)
          Product Details
          54-MSM3-P0
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1-2 µg/mL
          -
          Peptide Array (Array)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1, kappa
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          rACP5, 1070
          Immunogen
          Recombinant full-length human ACP5 protein
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          200 µg/mL
          Amount
          20 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4, with 0.05% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Immunohistochemistry (PFA fixed): incubate antibody for 30 min RT. Staining of formalin-fixed tissues requires heating tissue sections in 10mM Tris with 1mM EDTA, pH 9.0, for 45 min at 95 degrees C followed by cooling at RT for 20 minutes.
          Target Information
          Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher's disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin's lymphoma and in the sera of patients undergoing active bone turnover.
          Bioinformatics
          Protein Aliases: acid phosphatase type V; ACP5; human purple acid phosphatase; Tartrate-resistant acid ATPase; tartrate-resistant acid phosphatase 5a; tartrate-resistant acid phosphatase 5b; Tartrate-resistant acid phosphatase type 5; TR-AP; TRAPase; TrATPase; Type 5 acid phosphatase; unnamed protein product
          View more View less
          Gene Aliases: ACP5; HPAP; T5ap; TRACP; TRACP5a; TRACP5b; TRAP; TrATPase; TTRRAP
          View more View less
          UniProt ID: (Human) P13686, (Rat) P29288, (Mouse) Q05117
          View more View less
          Entrez Gene ID: (Human) 54, (Rat) 25732, (Mouse) 11433
          View more View less
          Function(s)
          acid phosphatase activity ferrous iron binding ferric iron binding hydrolase activity metal ion binding hydrolase phosphatase metabolite interconversion enzyme
          Process(es)
          nitric oxide biosynthetic process negative regulation of macrophage cytokine production response to lipopolysaccharide negative regulation of interleukin-1 beta production negative regulation of interleukin-12 production negative regulation of tumor necrosis factor production negative regulation of superoxide anion generation response to cytokine superoxide anion generation negative regulation of nitric oxide biosynthetic process bone resorption negative regulation of inflammatory response defense response to Gram-positive bacterium bone morphogenesis ossification negative regulation of cell adhesion metabolic process response to zinc ion response to organic cyclic compound dephosphorylation osteoclast differentiation positive regulation of cell migration multicellular organismal response to stress response to L-ascorbic acid response to cholesterol
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline