  • Applications
    • Bioprocesamiento
    • Cultivo celular y transfección
    • Terapia celular y génica
    • Cromatografía
    • Pruebas moleculares
    • Soluciones digitales
    • Extracción y análisis de ADN y ARN
    • Espectroscopía, análisis elemental y de isótopos
    • Ver todas las aplicaciones y técnicas
  • Servicios
    • Servicios 360° de CDMO y CRO
    • Servicios de CDMO
    • Servicios de CRO
    • Servicios personalizados
    • Servicios de leasing y financiación
    • Servicios de instrumentos
    • Informática de laboratorio
    • OEM y oferta comercial
    • Servicios de formación
    • Unity Lab Services
    • Ver todos los servicios
  • Ayuda y soporte técnico
    • Regístrese para obtener una cuenta
    • Cómo hacer un pedido
    • Asistencia para el instrumental
    • Centros de soporte técnico
    • Centros de formación
    • Vea todos los temas de ayuda y soporte técnico
      Cell Culture Plastics
  • A quiénes brindamos nuestros servicios
    • Industria biotecnológica
    • Sector biofarmacéutico
    • CDMO
    • Diagnósticos de laboratorio
    • Ciencias aplicadas e industriales
  • Promociones
  • Contacto
  • Pedido rápido
  • Estado del pedido y seguimiento
  • Documentos y certificados
Thermo Fisher Scientific Logo
Search
Buscar
Search button
          • Estado del pedido
          • Pedido rápido
            • Cuenta
            • Pedidos
            • Connect: laboratorio, datos, aplicaciones
            • Productos y proyectos personalizados
            • Services Central
          • Primary Antibodies ›
          • c-Met Antibodies
          Rockland
          Phospho-c-Met (Tyr1349, Tyr1356) Polyclonal Antibody
          View all (104) c-Met antibodies
          Tech Support
          Cite Phospho-c-Met (Tyr1349, Tyr1356) Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          Phospho-c-Met (Tyr1349, Tyr1356) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-c-Met (Tyr1349, Tyr1356) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          Phospho-c-Met (Tyr1349, Tyr1356) Antibody (600-401-989S) in WB
          Western blot using Rocklands affinity purified anti-c-Met pY1349pY1356 antibody shows detection of phosphorylated c-Met. Human mammary epithelial cells (B5/589) were serum deprived and stimulated with (+) and without (-) HGF. Cell lysates were immunoprecipitated using a human anti-c-Met antibody, resolved by SDS-PAGE, transferred to PVDF membrane and probed using various antibodies. Lane 1 and 2 where probe... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Phospho-c-Met (Tyr1349, Tyr1356) Antibody in Western Blot (WB)
          Phospho-c-Met (Tyr1349, Tyr1356) Antibody in Western Blot (WB)
          Phospho-c-Met (Tyr1349, Tyr1356) Polyclonal Antibody
          Product Details
          600-401-989S
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1 µg/mL
          -
          ELISA (ELISA)
          1:93,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding Y1349 and Y1356 of human c-Met protein.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.96 mg/mL
          Amount
          24 µg
          Purification
          Affinity chromatography
          Storage buffer
          0.02M potassium phosphate, pH 7.2, with 0.15M NaCl
          Contains
          0.01% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
          Antibody is specific for human c-Met protein phosphorylated at Y1349 and Y1356. A BLAST analysis was used to suggest cross-reactivity with c-Met from human, mouse and rat based on 100% homology with the immunizing sequence. Cross-reactivity with c-Met from other sources has not been determined.
          Target Information
          MET (cMET) is a receptor-like tyrosine kinase whose dysregulation has been linked to many types of human malignancies. After activation of the ligand, MET interacts with the PI3-kinase subunit PIK3R1, PLCG1, SRC, GRB2, and STAT3. There interactions lead to the activation of signaling cascades including RAS-ERK, PI3, kinase-AKT, and PLCgamma-PKC. MET plays a role in embryonic development including gastrulation, development of muscles and neurons, angiogenesis, and kidney formation. It also plays a role in adults including wound healing, organ regeneration, and tissue remodeling. MET has been linked to cancers including gastric, renal, and breast; therefore, making it a target for cancer therapeutics and diagnostic testing.
          Bioinformatics
          Protein Aliases: c-Met; cMet; EC 2.7.10.1; Hepatocyte growth factor receptor; HGF receptor; HGF-SF receptor; HGF/SF receptor; met oncogene; MET proto-oncogene protein; met proto-oncogene tyrosine kinase; oncogene MET; Proto-oncogene c-Met; sc MET; Scatter factor receptor; SF receptor; soluble c met; tyrosine kinase; hepatocyte growth factor receptor; HGFR; Tyrosine-protein kinase Met; unnamed protein product
          View more View less
          Gene Aliases: AI838057; AUTS9; c-Met; DA11; DFNB97; HGF; HGFR; MET; Par4; RCCP2
          View more View less
          UniProt ID: (Human) P08581, (Rat) P97523
          View more View less
          Entrez Gene ID: (Human) 4233, (Rat) 24553, (Mouse) 17295
          View more View less
          Function(s)
          nucleotide binding protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity hepatocyte growth factor-activated receptor activity protein binding ATP binding kinase activity transferase activity semaphorin receptor activity protein phosphatase binding identical protein binding molecular function activator activity beta-catenin binding protein complex binding phospholipase binding phosphatidylinositol 3-kinase binding protein heterodimerization activity transferase activity, transferring phosphorus-containing groups transmembrane signal receptor
          Process(es)
          endothelial cell morphogenesis liver development signal transduction cell surface receptor signaling pathway transmembrane receptor protein tyrosine kinase signaling pathway regulation of gene expression negative regulation of autophagy neuron differentiation pancreas development positive regulation of microtubule polymerization negative regulation of Rho protein signal transduction positive regulation of transcription from RNA polymerase II promoter hepatocyte growth factor receptor signaling pathway cell development animal organ development branching morphogenesis of an epithelial tube positive chemotaxis negative regulation of stress fiber assembly excitatory postsynaptic potential establishment of skin barrier negative regulation of thrombin receptor signaling pathway semaphorin-plexin signaling pathway negative regulation of hydrogen peroxide-mediated programmed cell death negative regulation of guanyl-nucleotide exchange factor activity positive regulation of endothelial cell chemotaxis negative regulation of transcription from RNA polymerase II promoter activation of MAPK activity neuron migration placenta development brain development muscle organ development skeletal muscle tissue development cell aging lactation male gonad development response to wounding response to acidic pH positive regulation of gene expression positive regulation of peptidyl-threonine phosphorylation negative regulation of peptidyl-threonine phosphorylation positive regulation of glucose transport positive regulation of neuron projection development oligodendrocyte development response to organic cyclic compound muscle cell migration myotube differentiation peptidyl-tyrosine phosphorylation central nervous system neuron differentiation sperm motility adult behavior positive regulation of peptidyl-serine phosphorylation response to drug glucose homeostasis response to peptide hormone positive regulation of DNA replication positive regulation of mitotic nuclear division protein autophosphorylation positive regulation of dendrite morphogenesis modulation of synaptic transmission myoblast proliferation regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling cellular response to inorganic substance cellular response to arsenic-containing substance cellular response to glucose stimulus cellular response to interleukin-6 cellular response to growth factor stimulus cellular response to peptide hormone stimulus protein phosphorylation phosphorylation
          Pedidos Plus Icon Minus Icon
          • Estado del pedido
          • Ayuda con el pedido
          • Pedido rápido
          • Centro de suministros
          • eProcurement (B2B)
          Asistencia Plus Icon Minus Icon
          • Ayuda y soporte técnico
          • Contacto
          • Centros de soporte técnico
          • Documentos y certificados
          • Informar de un problema del sitio web
          Recursos Plus Icon Minus Icon
          • Centros de formación
          • Promociones
          • Eventos y seminarios web
          • Redes sociales
          Acerca de Thermo Fisher Plus Icon Minus Icon
          • Acerca de nosotros Acerca de nosotros
          • Carreras profesionales Carreras profesionales
          • Inversores Inversores
          • Noticias Noticias
          • Responsabilidad social Responsabilidad social
          • Marcas registradas
          • Políticas y avisos
          Nuestra cartera Plus Icon Minus Icon
          Spain flag icon
          Spain
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline