    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • STAT2 Antibodies
          Rockland
          Phospho-STAT2 (Ser734) Polyclonal Antibody
          View all (34) STAT2 antibodies
          Tech Support
          Cite Phospho-STAT2 (Ser734) Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Phospho-STAT2 (Ser734) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-STAT2 (Ser734) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Phospho-STAT2 (Ser734) Antibody (600-401-H38S) in WB
          Western Blot of Rabbit anti-STAT2pS734 antibody. Lane 1: U6A cells (STAT2 deficient) untreated. Lane 2: U6A cells reconstituted with STAT2 untreated. Lane 3: U6A cells, STAT2 treated with IFN alpha(1h). Lane 4: U6A cells, STAT2 + IFN alpha (4h). Lane 5: U6A cells, STAT2 + IFN alpha (8h). Lane 6: U6A cells reconstituted with S734A-STAT2 mutant untreated. Lane 7: U6A cells, S734A-STAT2 treated with IFN alph... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Phospho-STAT2 (Ser734) Antibody in Western Blot (WB)
          Phospho-STAT2 (Ser734) Polyclonal Antibody
          Product Details
          600-401-H38S
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          ELISA (ELISA)
          1:5,000
          -
          Product Specifications
          Species Reactivity
          Human, Non-human primate
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          STAT2 pS734 antibody was prepared from whole rabbit serum produced by repeated immunizations with a phosphorylated synthetic peptide corresponding to serine 734 a region near the C-terminus of human STAT2 protein.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.64 mg/mL
          Amount
          16 µg
          Purification
          Affinity chromatography
          Storage buffer
          0.02M potassium phosphate, pH 7.2, with 0.15M NaCl
          Contains
          0.01% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
          STAT2 pS734 affinity purified antibody is directed against human STAT2 protein. This antibody is specific for phosphorylated S734 human STAT2. A BLAST analysis was used to suggest cross-reactivity with STAT2 pS734 protein from human and monkey sources based on homology with the immunizing sequence. Reactivity against homologues from other sources is not known.
          Target Information
          STAT2 is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. In response to interferon (IFN), this protein forms a complex with STAT1 and IFN regulatory factor family protein p48 (ISGF3G), in which this protein acts as a transactivator, but lacks the ability to bind DNA directly. Transcription adaptor P300/CBP (EP300/CREBBP) has been shown to interact specifically with this protein, which is thought to be involved in the process of blocking IFN-alpha response by adenovirus.
          Bioinformatics
          Protein Aliases: interferon alpha induced transcriptional activator; MGC59816; p113; Signal transducer and activator of transcription 2; signal transducer and activator of transcription 2, 113kDa; STAT; STAT 2; STAT-2; unnamed protein product
          View more View less
          Gene Aliases: IMD44; ISGF-3; P113; PTORCH3; STAT113; STAT2
          View more View less
          UniProt ID: (Human) P52630
          View more View less
          Entrez Gene ID: (Human) 6773
          View more View less
          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding identical protein binding ubiquitin-like protein ligase binding
          Process(es)
          regulation of protein phosphorylation regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter defense response signal transduction JAK-STAT cascade regulation of cell proliferation response to peptide hormone positive regulation of transcription from RNA polymerase II promoter defense response to virus type I interferon signaling pathway negative regulation of type I interferon-mediated signaling pathway regulation of mitochondrial fission
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline