    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • TAFA2 Antibodies
          AbboMax
          TAFA2 Polyclonal Antibody
          View all (5) TAFA2 antibodies
          Tech Support
          Cite TAFA2 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          TAFA2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          TAFA2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          TAFA2 Antibody (600-460) in IHC (P)
          Immunohistochemistry: Human tonsil tissue (FFPE) was immune-stained with Anti-TAFA2 antibody, (Cat# 600-460) at 1:100 for 10 min @ RT. Staining of formalin-fixed tissue requires boiling tissue sections in 10 mM Citrate Buffer, pH 6.0 for 10 min followed by cooling at RT for 20 min. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TAFA2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TAFA2 Antibody in Western Blot (WB)
          TAFA2 Polyclonal Antibody
          Product Details
          600-460
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.1-1 µg/mL
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          2-10 µg/mL
          -
          ELISA (ELISA)
          0.01-0.1 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          A synthetic peptide corresponding to the N-terminus of TAFA2 protein
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS with proprietary stabilizer
          Contains
          0.01% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Positive control: Brain tissue
          Cellular location: Extracellular or cytoplasmic.
          Target Information
          This gene is a member of the TAFA family which is composed of five highly homologous genes that encode small secreted proteins. These proteins contain conserved cysteine residues at fixed positions, and are distantly related to MIP-1alpha, a member of the CC-chemokine family. The TAFA proteins are predominantly expressed in specific regions of the brain, and are postulated to function as brain-specific chemokines or neurokines, that act as regulators of immune and nervous cells.
          Bioinformatics
          Protein Aliases: Chemokine-like protein TAFA-2; family with sequence similarity 19 (chemokine (C-C motif)-like), member A2; family with sequence similarity 19 member A2, C-C motif chemokine like; protein FAM19A2; unnamed protein product
          View more View less
          Gene Aliases: 6330575M02; FAM19A2; FAM19A2 TAFA-2; TAFA-2; TAFA2
          View more View less
          UniProt ID: (Mouse) Q7TPG7, (Human) Q8N3H0
          View more View less
          Entrez Gene ID: (Mouse) 268354, (Rat) 680647, (Human) 338811
          View more View less
          Function(s)
          molecular_function receptor agonist activity chemokine
          Process(es)
          biological_process signal transduction memory visual learning
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline