    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • CD3g Antibodies
          Proteintech
          CD3 gamma Monoclonal Antibody (2E9G7)
          1 Reference
          View all (14) CD3g antibodies
          Tech Support
          Cite CD3 gamma Monoclonal Antibody (2E9G7)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          CD3 gamma Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          CD3 gamma Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          CD3 gamma Antibody (60347-1-IG) in IHC (P)
          Immunohistochemistry of paraffin-embedded human tonsillitis tissue slide using 60347-1-IG ( CD3 gamma antibody at dilution of 1:400 (under 40x lens). heat mediated antigen retrieved with Sodium Citrate buffer (pH6). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD3 gamma Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD3 gamma Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD3 gamma Antibody in Western Blot (WB)
          CD3 gamma Antibody in Flow Cytometry (Flow)
          CD3 gamma Monoclonal Antibody (2E9G7)
          Product Details
          60347-1-IG
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:8,000
          -
          Immunohistochemistry (IHC)
          -
          View 1 publication 1 publication
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.2 µg/1x10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2E9G7
          Immunogen
          CD3 gamma Fusion Protein Ag15502 (23-182 aa encoded by BC113830)
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite 594
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          150 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice (domestic); Dry ice (international)
          Product Specific Information
          Immunogen sequence: QSIKGNHLV KVYDYQEDGS VLLTCDAEAK NITWFKDGKM IGFLTEDKKK WNLGSNAKDP RGMYQCKGSQ NKSKPLQVYY RMCQNCIELN AATISGFLFA EIVSIFVLAV GVYFIAGQDG VRQSRASDKQ TLLPNDQLYQ PLKDREDDQY SHLQGNQLRR N (23-182 aa encoded by BC113830)
          Target Information
          CD3G is the CD3-gamma polypeptide, which together with CD3-epsilon, -delta and -zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T-cell receptor-CD3 complex. This complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. The genes encoding the epsilon, gamma and delta polypeptides are located in the same cluster on chromosome 11. Defects in the CD3G gene are associated with T cell immunodeficiency.
          Bioinformatics
          Protein Aliases: CD3g; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); T-cell antigen receptor complex, gamma subunit of T3; T-cell receptor T3 gamma chain; T-cell surface glycoprotein CD3 gamma chain; unnamed protein product
          View more View less
          Gene Aliases: CD3-GAMMA; CD3G; CD3GAMMA; IMD17; T3G
          View more View less
          UniProt ID: (Human) P09693
          View more View less
          Entrez Gene ID: (Human) 917
          View more View less
          Function(s)
          transmembrane signaling receptor activity protein binding receptor signaling complex scaffold activity T cell receptor binding identical protein binding
          Process(es)
          adaptive immune response immune system process establishment or maintenance of cell polarity cell surface receptor signaling pathway protein transport T cell activation positive thymic T cell selection gamma-delta T cell activation alpha-beta T cell activation T cell receptor signaling pathway protein-containing complex assembly regulation of lymphocyte apoptotic process
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline