    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • ICOS (CD278) Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Invitrogen
          CD278 (ICOS) Monoclonal Antibody (7E.17G9), Super Bright™ 436, eBioscience™
          2 Published Figures
          3 References
          View all (98) ICOS (CD278) antibodies
          Tech Support
          Cite CD278 (ICOS) Monoclonal Antibody (7E.17G9), Super Bright™ 436, eBioscience™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          • Published Figures (2)
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          CD278 (ICOS) Antibody (62-9942-80) in Flow
          BALB/c mouse splenocytes were unstimulated (left) or stimulated for 72 hours with Con A (Product # 00-4978-03) (right). Cells were then stained with CD25 Monoclonal Antibody, Alexa Fluor 488 (Product # 53-0251-82) and 0.25 µg of CD278 Monoclonal Antibody, Super Bright 436. Total viable cells were used for analysis, as determined by Fixable Viability Dye eFluor 660 (Product # 65-0864-18). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          Product Details
          62-9942-80
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          0.5 µg/test
          View 3 publications 3 publications
          Product Specifications
          Host/Isotype
          Rat / IgG2b, kappa
          Recommended Isotype Control
          Rat IgG2b kappa Isotype Control (eB149/10H5), Super Bright™ 436, eBioscience™
          Class
          Monoclonal
          Type
          Antibody
          Clone
          7E.17G9
          Conjugate
          Super Bright™ 436 Super Bright™ 436 Super Bright™ 436
          • Unconjugated
          • APC 
          • Biotin 
          • FITC
          • Functional Grade
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PerCP-eFluor 710
          • Super Bright 645
          • Request custom conjugation
          Excitation/Emission Max
          413/431 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.2 mg/mL
          Amount
          25 µg
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Wet ice
          RRID
          AB_2744817
          Product Specific Information
          Description: The 7E.17G9 monoclonal antibody reacts with mouse ICOS (Inducible COStimulatory molecule), a T cell specific molecule and a third member of the CD28/CTLA-4 family. A homodimer of 47-57 kDa, ICOS is expressed on activated T cells, has potent costimulatory activity for T cell activation and proliferation and is required for humoral immune response. ICOS binds to its ligand on activated APC including B cells called B7h/B7RP-1 and is thought to play a protective role in inflammatory autoimmune diseases. ICOS may be involved in the development of Th2 cells.
          Applications Reported: This 7E.17G9 antibody has been reported for use in flow cytometric analysis.
          Applications Tested: This 7E.17G9 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This may be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
          Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.
          When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information.
          Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser
          Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
          Target Information
          ICOS (CD278) encodes a protein that is a member of the CD28 and CTLA-4 cell-surface receptor family. It functions as a homodimer and has a crucial role in cell-cell signaling, immune responses, and the regulation of cell proliferation. ICOS has been linked to various disorders, including Immunodeficiency, Common Variable, 1 and Common Variable Immunodeficiency.
          How to use the Panel Builder
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline