    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
          • Primary Antibodies ›
          • EBAG9 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Proteintech
          RCAS1 Monoclonal Antibody (4H8A12)
          2 References
          View all (24) EBAG9 antibodies
          Tech Support
          Cite RCAS1 Monoclonal Antibody (4H8A12)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (9)
          RCAS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          RCAS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 9
          RCAS1 Antibody (66170-1-IG) in IHC (P)
          Immunohistochemical analysis of paraffin-embedded Human breast cancer tissue slide using (Product # 66170-1-Ig) RCAS1 antibody at dilution of 1:200 (under 40x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          RCAS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RCAS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RCAS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RCAS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RCAS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RCAS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RCAS1 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          RCAS1 Antibody in Western Blot (WB)
          RCAS1 Antibody in Flow Cytometry (Flow)
          RCAS1 Monoclonal Antibody (4H8A12)
          Product Details
          66170-1-IG
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:2,000
          View 1 publication 1 publication
          Immunohistochemistry (Paraffin) (IHC (P))
          1:20-1:200
          -
          Immunohistochemistry (PFA fixed) (IHC (PFA))
          1:400
          -
          Immunocytochemistry (ICC/IF)
          -
          View 1 publication 1 publication
          Flow Cytometry (Flow)
          Assay-dependent
          -
          Product Specifications
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          4H8A12
          Immunogen
          RCAS1 Fusion Protein Ag2905 (1-213 aa encoded by BC017729)
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite 594
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          0.67 mg/mL
          Amount
          100.5 µg
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice (domestic); Dry ice (international)
          Product Specific Information
          Immunogen sequence: MAITQFRLF KFCTCLATVF SFLKRLICRS GRGRKLSGDQ ITLPTTVDYS SVPKQTDVEE WTSWDEDAPT SVKIEGGNGN VATQQNSLEQ LEPDYFKDMT PTIRKTQKIV IKKREPLNFG IPDGSTGFSS RLAATQDLPF IHQSSELGDL DTWQENTNAW EEEEDAAWQA EEVLRQQKLA DREKRAAEQQ RKKMEKEAQR LMKKEQNKIG VKLS (1-213 aa encoded by BC017729)
          Target Information
          This gene was identified as an estrogen-responsive gene. Regulation of transcription by estrogen is mediated by estrogen receptor which binds to the estrogen-responsive element found in the 5'-flanking region of this gene. The encoded protein is a tumor-associated antigen that is expressed at high frequency in a variety of cancers. Two transcript variants differing in the 5' UTR, but encoding the same protein, have been identified for this gene.
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline