    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • HDDC2 Antibodies
          Proteintech
          HDDC2 Monoclonal Antibody (2E1B2)
          View all (3) HDDC2 antibodies
          Tech Support
          Cite HDDC2 Monoclonal Antibody (2E1B2)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          HDDC2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          HDDC2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          HDDC2 Antibody (68391-1-IG) in ICC/IF
          Immunofluorescent analysis of (-20°C Methanol) fixed Saos-2 cells using HDDC2 antibody (68391-1-Ig, Clone: 2E1B2) at dilution of 1:400 and CoraLite®488-Conjugated AffiniPure Goat Anti-Mouse IgG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          HDDC2 Antibody in Immunocytochemistry (ICC/IF)
          HDDC2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HDDC2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HDDC2 Antibody in Western Blot (WB)
          HDDC2 Antibody in Flow Cytometry (Flow)
          HDDC2 Monoclonal Antibody (2E1B2)
          Product Details
          68391-1-IG
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:500-1:2,000
          -
          Immunocytochemistry (ICC/IF)
          1:200-1:800
          -
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2E1B2
          Immunogen
          HDDC2 fusion protein Ag33258
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          150 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MASVSSATFS GHGARSLLQF LRLVGQLKRV PRTGWVYRNV QRPESVSDHM YRMAVMAMVI KDDRLNKDRC VRLALVHDMA ECIVGDIAPA DNIPKEEKHR REEEAMKQIT QLLPEDLRKE LYELWEEYET QSSAEAKFVK QLDQCEMILQ ASEYEDLEHK PGRLQDFYDS TAGKFNHPEI VQLVSELEAE RSTNIAAAAS EPHS
          Target Information
          Catalyzes the dephosphorylation of the nucleoside 5'-monophosphates deoxyadenosine monophosphate (dAMP), deoxycytidine monophosphate (dCMP), deoxyguanosine monophosphate (dGMP) and deoxythymidine monophosphate (dTMP). [UniProt]
          Bioinformatics
          Protein Aliases: 5'-deoxynucleotidase HDDC2; HCV NS5A-transactivated protein 2; HD domain-containing protein 2; Hepatitis C virus NS5A-transactivated protein 2; testicular tissue protein Li 83
          View more View less
          Gene Aliases: 2310057G13Rik; C6orf74; CGI-130; dJ167O5.2; HDDC2; NS5ATP2; RGD1307562
          View more View less
          UniProt ID: (Human) Q7Z4H3, (Mouse) Q3SXD3
          View more View less
          Entrez Gene ID: (Human) 51020, (Mouse) 69692, (Rat) 361462
          View more View less
          Function(s)
          5'-deoxynucleotidase activity protein binding hydrolase activity metal ion binding molecular_function
          Process(es)
          biological_process deoxyribonucleoside monophosphate catabolic process
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline