    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SYAP1 Antibodies
          Proteintech
          SYAP1 Monoclonal Antibody (2E12A5)
          View all (10) SYAP1 antibodies
          Tech Support
          Cite SYAP1 Monoclonal Antibody (2E12A5)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          SYAP1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SYAP1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          SYAP1 Antibody (68410-1-IG) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using SYAP1 antibody (68410-1-Ig, Clone: 2E12A5) at dilution of 1:800 and CoraLite®488-Conjugated Goat Anti-Mouse IgG (H+L), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SYAP1 Antibody in Immunocytochemistry (ICC/IF)
          SYAP1 Antibody in Western Blot (WB)
          SYAP1 Antibody in Immunoprecipitation (IP)
          SYAP1 Monoclonal Antibody (2E12A5)
          Product Details
          68410-1-IG
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunocytochemistry (ICC/IF)
          1:400-1:1,600
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          0.5-4.0 µg for 1.0-3.0 mg of total protein lysate
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2E12A5
          Immunogen
          SYAP1 fusion protein Ag9626
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          150 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MFRGLSSWLG LQQPVAGGGQ PNGDALPEQP SETVAESAEE ELQQAGDQEL LHQAKDFGNY LFNFASAATK KITESVAETA QTIKKSVEEG KIDGIIDKTI IGDFQKEQKK FVEEQHTKKS EAAVPPWVDT NDEETIQQQI LALSADKRNF LRDPPAGVQF NFDFDQMYPV ALVMLQEDEL LSKMRFALVP KLVKEEVFWR NYFYRVSLIK QSAQLTALAA QQQAAGKEEK SNGREQDLPL AEAVRPKTPP VVIKSQLKTQ EDEEEISTSP GVSEFVSDAF DACNLNQEDL RKEMEQLVLD KKQEETAVLE EDSADWEKEL QQELQEYEVV TESEKRDENW DKEIEKMLQE EN
          Target Information
          SYAP1 plays a role in adipocyte differentiation by promoting mTORC2-mediated phosphorylation of AKT1 at 'Ser-473' after growth factor stimulation.
          Bioinformatics
          Protein Aliases: BSD domain-containing signal transducer and Akt interactor protein; BSTA; SAP47 homolog; synapse associated protein 1, SAP47 homolog; Synapse-associated protein 1; unnamed protein product
          View more View less
          Gene Aliases: 2010110O17Rik; AW011796; BSTA; PRO3113; SYAP1
          View more View less
          UniProt ID: (Human) Q96A49, (Mouse) Q9D5V6
          View more View less
          Entrez Gene ID: (Human) 94056, (Mouse) 67043, (Rat) 302678
          View more View less
          Function(s)
          protein binding molecular_function
          Process(es)
          biological_process cell differentiation cellular response to insulin stimulus cellular response to platelet-derived growth factor stimulus TORC2 signaling positive regulation of fat cell differentiation regulation of short-term neuronal synaptic plasticity cellular response to epidermal growth factor stimulus positive regulation of protein serine/threonine kinase activity cellular response to insulin-like growth factor stimulus
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline