    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • S6 Antibodies
          This clone is available in multiple conjugated formats in both full and sample sizes. For more options, search using the clone ID "cupk43k" or explore the "Additional formats" section on this page.
          Invitrogen
          Phospho-S6 (Ser235, Ser236) Recombinant Mouse Monoclonal Antibody (cupk43k), Alexa Fluor™ Plus 555
          Advanced Verification
          View all (148) S6 antibodies
          Tech Support
          Cite Phospho-S6 (Ser235, Ser236) Recombinant Mouse Monoclonal Antibody (cupk43k), Alexa Fluor™ Plus 555
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (7)
          • Advanced Verification (2)
          Phospho-S6 (Ser235, Ser236) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-S6 (Ser235, Ser236) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 9
          Phospho-S6 (Ser235, Ser236) Antibody (740057MP555) in ICC/IF
          Immunofluorescence analysis of Phospho-S6 (Ser235, Ser236) was performed using Phospho-S6 (Ser235, Ser236) Recombinant Mouse Monoclonal Antibody (cupk43k), Alexa Fluor™ Plus 555 (Product # 740057MP555) on U-2OS cells treated with 50 nM Rapamycin for 4 hr followed by Rec Hu IGF-1 50 ng/mL for 6 hr. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton X-100 for 15 minut... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Phospho-S6 (Ser235, Ser236) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-S6 (Ser235, Ser236) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-S6 (Ser235, Ser236) Antibody in Western Blot (WB)
          Phospho-S6 (Ser235, Ser236) Antibody in Western Blot (WB)
          Phospho-S6 (Ser235, Ser236) Antibody in Western Blot (WB)
          Phospho-S6 (Ser235, Ser236) Antibody in Flow Cytometry (Flow)
          Phospho-S6 (Ser235, Ser236) Antibody in Flow Cytometry (Flow)
          Phospho-S6 (Ser235, Ser236) Antibody
          Phospho-S6 (Ser235, Ser236) Antibody
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          740057MP555
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:400-1:1,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50
          -
          Immunocytochemistry (ICC/IF)
          1:500
          -
          Flow Cytometry (Flow)
          0.25-0.5 µg/test
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1, kappa
          Expression System
          Expi293
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          cupk43k
          Conjugate
          Alexa Fluor™ Plus 555 Alexa Fluor™ Plus 555 Alexa Fluor™ Plus 555
          • Unconjugated
          • Alexa Fluor Plus 488 
          • Alexa Fluor Plus 647 
          • Request custom conjugation
          Excitation/Emission Max
          558/572 nm View spectra spectra
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          50 µg
          Purification
          Protein A/G
          Storage buffer
          proprietary buffer, pH 6.3-7.3
          Contains
          0.008% Bromonitrodioxane, 0.008% Methylisothiazolone
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Wet ice
          RRID
          AB_3699410
          Product Specific Information
          Alexa Fluor™ Plus recombinant antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning data. Alexa Fluor™ Plus antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor™ Plus antibodies provide brighter signal compared to leading Alexa Fluor™ antibodies, providing you with better signal-to-noise for your critical experiments. These antibodies show better specificity and lot-to-lot consistency as these are recombinant antibodies, generated by cloning specific genes for the desired antibodies into an expression vector and expressed in vitro.
          Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining.
          It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
          Excitation: 553 nm; Emission: 568 nm; Laser: Yellow Laser
          Filtration: 0.2 µm post-manufacturing filtered.
          Target Information
          Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a cytoplasmic ribosomal protein that is a component of the 40S subunit. The protein belongs to the S6E family of ribosomal proteins. It is the major substrate of protein kinases in the ribosome, with subsets of five C-terminal serine residues phosphorylated by different protein kinases. Phosphorylation is induced by a wide range of stimuli, including growth factors, tumor-promoting agents, and mitogens. Dephosphorylation occurs at growth arrest. The protein may contribute to the control of cell growth and proliferation through the selective translation of particular classes of mRNA. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
          Bioinformatics
          Protein Aliases: 40S ribosomal protein S6; OTTHUMP00000021121; OTTHUMP00000021122; phosphoprotein NP33; ribosomal protein S6; RP11-513M16.6; RS6; unnamed protein product
          View more View less
          Gene Aliases: eS6; S6
          View more View less
          UniProt ID: (Human) P62753
          View more View less
          Entrez Gene ID: (Human) 6194
          View more View less
          Function(s)
          RNA binding structural constituent of ribosome protein binding protein kinase binding
          Process(es)
          cytoplasmic translation rRNA processing translation positive regulation of cell proliferation TOR signaling ribosomal small subunit biogenesis glucose homeostasis positive regulation of apoptotic process
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-green-5d6d8bc979-v94d6:80/100.66.72.150:80.
          git-commit: e71199a02a6c1449b2ac9ded8316d5734e8aa2de
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.62.0-2026.11.45-1.0