    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • Neurofascin Antibodies
          Antibodies Inc
          Pan-Neurofascin Monoclonal Antibody (L11A/41)
          View all (16) Neurofascin antibodies
          Tech Support
          Cite Pan-Neurofascin Monoclonal Antibody (L11A/41)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Pan-Neurofascin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Pan-Neurofascin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Pan-Neurofascin Antibody (75-027) in WB
          Adult rat brain membrane immunoblot. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Pan-Neurofascin Antibody in Western Blot (WB)
          Pan-Neurofascin Monoclonal Antibody (L11A/41)
          Product Details
          75-027
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          5 µg
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          L11A/41
          Immunogen
          Fusion protein 1066-1174 (intracellular domain common to NF-155 and NF-186) of rat NF-155 (accession number AAL27854) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4
          Contains
          0.065% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Neurofascin encodes an L1 family immunoglobulin cell adhesion molecule with multiple IGcam and fibronectin domains. The protein functions in neurite outgrowth, neurite fasciculation, and organization of the axon initial segment (AIS) and nodes of Ranvier on axons during early development. Both the AIS and nodes of Ranvier contain high densities of voltage-gated Na+ (Nav) channels which are clustered by interactions with cytoskeletal and scaffolding proteins including this protein, gliomedin, ankyrin 3 (ankyrin-G), and betaIV spectrin. This protein links the AIS extracellular matrix to the intracellular cytoskeleton. Neurofascin undergoes extensive alternative splicing, and the full-length nature of some variants has not been determined.
          Bioinformatics
          Protein Aliases: DKFZp686P2250; KIAA0756FLJ46866; Neurofascin; neurofascin homolog; unnamed protein product
          View more View less
          Gene Aliases: KIAA0756; NEDCPMD; NF; NFASC; NRCAML
          View more View less
          UniProt ID: (Human) O94856, (Rat) P97685
          View more View less
          Entrez Gene ID: (Human) 23114, (Rat) 116690
          View more View less
          Function(s)
          protein binding protein binding involved in cell-cell adhesion protein domain specific binding protein binding involved in heterotypic cell-cell adhesion scaffold/adaptor protein
          Process(es)
          cell adhesion axon guidance brain development peripheral nervous system development myelination cell-cell adhesion protein localization to paranode region of axon transmission of nerve impulse paranodal junction assembly heterotypic cell-cell adhesion clustering of voltage-gated sodium channels synapse organization protein localization to juxtaparanode region of axon protein targeting to plasma membrane
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0