    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • KV3.1b (KCNC1) Antibodies
          Antibodies Inc
          Kv3.1B K+ Channel Monoclonal Antibody (N16B/8), FL650
          View all (6) KV3.1b (KCNC1) antibodies
          Tech Support
          Cite Kv3.1B K+ Channel Monoclonal Antibody (N16B/8), FL650
          Additional Information:
          {{banner.description}}
          Kv3.1B K+ Channel Monoclonal Antibody (N16B/8), FL650
          Product Details
          75-041-FL650
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          1:100
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Hamster, Human, Mouse, Non-human primate, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N16B/8
          Immunogen
          Fusion protein amino acids 437-585 (cytoplasmic C-terminus) of rat Kv3.1b (accession number P25122) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Voltage-gated K+ channels are important determinants of neuronal membrane excitability. Moreover, differences in K+ channel expression patterns and densities contribute to the variations in action potential waveforms and repetitive firing patterns evident in different neuronal cell types (Maletic-Savatic et al., 1995; Pongs, 1999; Blaine and Ribera, 1998; Burger and Ribera, 1996). The Kv3.1 potassium channel is expressed at high levels in neurons that characteristically fire rapid trains of action potentials (Gan et al., 1999). Particularly high levels of this channel are found in neurons of the auditory brainstem. These neurons appear to participate in neural circuits that determine the intensity and timing of auditory stimuli and use this information to determine the location of sounds in space (von Hehn et al., 2004).
          Bioinformatics
          Protein Aliases: NGK2; Potassium channel gene 1 (alternative splicing product described in Luneau et al 1991); potassium channel, voltage gated Shaw related subfamily C, member 1; potassium voltage gated channel, Shaw-related subfamily, member 1; Potassium voltage-gated channel subfamily C member 1; potassium voltage-gated channel, Shaw-related subfamily, member 1; RAW2; Voltage-gated potassium channel KCNC1; voltage-gated potassium channel protein KV3.1; Voltage-gated potassium channel subunit Kv3.1; Voltage-gated potassium channel subunit Kv4
          View more View less
          Gene Aliases: C230009H10Rik; EPM7; I79_017349; KCNC1; Kcr2-1; KShIIIB; KV3.1; KV4; NGK2; NGK2-KV4; Shaw
          View more View less
          UniProt ID: (Human) P48547, (Mouse) P15388, (Rat) P25122
          View more View less
          Entrez Gene ID: (Human) 3746, (Chinese hamster) 100763523, (Mouse) 16502, (Rat) 25327
          View more View less
          Function(s)
          ion channel activity voltage-gated potassium channel activity delayed rectifier potassium channel activity potassium channel activity protein binding kinesin binding gated channel activity ion channel binding voltage-gated monoatomic ion channel activity involved in regulation of presynaptic membrane potential voltage-gated ion channel activity voltage-gated ion channel
          Process(es)
          action potential ion transport potassium ion transport response to toxic substance response to light intensity response to auditory stimulus response to amine cerebellum development optic nerve development globus pallidus development corpus callosum development ion transmembrane transport regulation of ion transmembrane transport positive regulation of ion transmembrane transport response to potassium ion protein homooligomerization protein tetramerization transmembrane transport cellular response to xenobiotic stimulus response to fibroblast growth factor potassium ion transmembrane transport regulation of presynaptic membrane potential regulation of potassium ion transmembrane transport positive regulation of potassium ion transmembrane transport response to nerve growth factor transport brain development cellular response to drug
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline