    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SH3GL1 Antibodies
          Antibodies Inc
          SH3GL1/Endophilin A2 Monoclonal Antibody (K51/1), FL650
          View all (29) SH3GL1 antibodies
          Tech Support
          Cite SH3GL1/Endophilin A2 Monoclonal Antibody (K51/1), FL650
          Additional Information:
          {{banner.description}}
          SH3GL1/Endophilin A2 Monoclonal Antibody (K51/1), FL650
          Product Details
          75-049-FL650
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          K51/1
          Immunogen
          Synthetic peptide amino acids 287-302 of rat SH3GL1 (accession number AAH70893).
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          The protein encoded by this gene contains a SH2 domain and a SOCS BOX domain. The protein thus belongs to the suppressor of cytokine signaling (SOCS), also known as STAT-induced STAT inhibitor (SSI), protein family. SOCS family members are known to be cytokine-inducible negative regulators of cytokine signaling. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
          Bioinformatics
          Protein Aliases: endophilin II; Endophilin-2; Endophilin-A2; MGC111371; SH3 domain protein 2B; SH3 domain-containing GRB2-like protein 1; SH3p8
          View more View less
          Gene Aliases: C77078; EEN; Sh3d2b; Sh3gl1; SH3P8
          View more View less
          UniProt ID: (Mouse) Q62419, (Rat) O35964
          View more View less
          Entrez Gene ID: (Mouse) 20405, (Rat) 81922
          View more View less
          Function(s)
          protein binding lipid binding phosphatase binding identical protein binding GTPase binding
          Process(es)
          endocytosis biological_process
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline