    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • KCNT2 Antibodies
          Antibodies Inc
          Slo2.1/Slick K+ Channel Monoclonal Antibody (N11/33)
          View all (10) KCNT2 antibodies
          Tech Support
          Cite Slo2.1/Slick K+ Channel Monoclonal Antibody (N11/33)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          Slo2.1/Slick K+ Channel Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          Slo2.1/Slick K+ Channel Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          Slo2.1/Slick K+ Channel Antibody (75-055) in IHC (F)
          Adult rat whole brain immunohistochemistry. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Slo2.1/Slick K+ Channel Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Slo2.1/Slick K+ Channel Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Slo2.1/Slick K+ Channel Monoclonal Antibody (N11/33)
          Product Details
          75-055
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500
          -
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          1:100
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          5 µg
          -
          Immunomicroscopy (IM)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat, Xenopus
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N11/33
          Immunogen
          Fusion protein amino acids 564-624 of mouse Slo2.1 (accession number XP_920840) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4
          Contains
          0.065% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Kcnt2 calcium (Ca2+) -activated potassium channels (KCa) are a group of 6/7-TM ion channels that selectively transport K+ ions across biological membranes. KCa channels are broadly classified into three subtypes: SK, IK and BK channels (small, intermediate and big conductance). Kcnt2 is an outward rectifying potassium channel, and produces rapidly activating outward rectifier K(+) currents. Kcnt2 is activated by high intracellular sodium and chloride levels. Further, Kcnt2 channel activity is inhibited by ATP and by inhalation anesthetics, such as isoflurane. Functionally, Kcnt2 is inhibited upon stimulation of G-protein coupled receptors, such as CHRM1 and GRIA1.
          Bioinformatics
          Protein Aliases: Potassium channel subfamily T member 2; potassium channel, subfamily T, member 2; Sequence like an intermediate conductance potassium channel subunit; Sodium and chloride-activated ATP-sensitive potassium channel Slo2.1; sodium- and chloride-activated ATP-sensitive potassium channel; sodium- and chloride-activated ATP-sensitive potassium channel subunit
          View more View less
          Gene Aliases: E330038N15Rik; Kcnt2; Slick; Slo2.2
          View more View less
          UniProt ID: (Rat) Q6UVM4, (Mouse) D3Z649
          View more View less
          Entrez Gene ID: (Rat) 304827, (Mouse) 240776
          View more View less
          Function(s)
          intracellular sodium activated potassium channel activity voltage-gated potassium channel activity ATP binding calcium-activated potassium channel activity potassium channel activity ion channel
          Process(es)
          potassium ion transmembrane transport potassium ion transport regulation of ion transmembrane transport
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline