  • Aplicaciones
    • Cultivo celular y transfección
    • Citometría de flujo
    • Investigación sobre el cáncer
    • Cromatografía
    • Secuenciación
    • PCR
    • Soluciones para laboratorio
    • Diagnóstico de alergias
    • Ver todas las aplicaciones y técnicas
  • Servicios
    • Servicios Personalizados
    • Servicios de Capacitación
    • Informática para laboratorios de ámbito empresarial
    • Servicios financieros y de arrendamiento
    • Servicios 360° de CDMO y CRO
    • Servicios de CDMO
    • Servicios de CRO
    • Ver todos los servicios
  • Ayuda y Soporte
    • Crear una nueva cuenta
    • Cómo hacer el pedido
    • Póngase en contacto con nosotros
    • Cambio de ubicación
    • Ver toda la ayuda y soporte técnico
      Cell Culture Plastics
  • A quiénes brindamos nuestros servicios
    • Industria biotecnológica
    • Sector biofarmacéutico
    • CDMO
    • Diagnósticos de laboratorio
    • Ciencias aplicadas e industriales
  • Ofertas especiales
  • Contáctenos
  • Orden Rápida
  • Documentos y certificados
Thermo Fisher Scientific Logo
Search
Buscar
Search button
          • Contáctenos
          • Orden Rápida
          • Iniciar sesión
            Iniciar sesión
            ¿No tiene una cuenta? Crear una cuenta
            • Cuenta
            • Pedidos
            • Productos y proyectos personalizados
          • Primary Antibodies ›
          • Kir2.3 (KCNJ4) Antibodies
          Antibodies Inc
          Kir2.3 K+ Channel Monoclonal Antibody (N25/35), FL490
          View all (9) Kir2.3 (KCNJ4) antibodies
          Tech Support
          Cite Kir2.3 K+ Channel Monoclonal Antibody (N25/35), FL490
          Additional Information:
          {{banner.description}}
          Kir2.3 K+ Channel Monoclonal Antibody (N25/35), FL490
          Product Details
          75-069-FL490
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N25/35
          Immunogen
          Fusion protein amino acids 390-445 of human Kir2.3 (accession number NP_032453) produced recombinantly in E. Coli.
          View immunogen
          Conjugate
          FL490 FL490 FL490
          • Unconjugated
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          491/515 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Several different potassium channels are known to be involved with electrical signaling in the nervous system. One class is activated by depolarization whereas a second class is not. The latter are referred to as inwardly rectifying K+ channels, and they have a greater tendency to allow potassium to flow into the cell rather than out of it. This asymmetry in potassium ion conductance plays a key role in the excitability of muscle cells and neurons. The protein encoded by this gene is an integral membrane protein and member of the inward rectifier potassium channel family. The encoded protein has a small unitary conductance compared to other members of this protein family. Two transcript variants encoding the same protein have been found for this gene.
          Bioinformatics
          Protein Aliases: BIR11; Brain inwardly rectifying K(+) channel 2; Hippocampal inward rectifier; hippocampal inward rectifier potassium channel; HIR; HIRK2; HRK1; Inward rectifier K(+) channel Kir2.3; inward rectifier potassium channel; Inward rectifier potassium channel 4; IRK-3; IRK3; KCNJ4; Kir 2.3; Potassium channel, inwardly rectifying subfamily J member 4; potassium channel, inwardly rectifying subfamily J, member 4; potassium inwardly-rectifying channel, subfamily J, member 4; potassium voltage-gated channel subfamily J member 4
          View more View less
          Gene Aliases: HIR; HIRK2; HRK1; IRK-3; IRK3; Kcnf2; KCNJ4; Kir2.3; MB-IRK3
          View more View less
          UniProt ID: (Human) P48050, (Rat) P52190
          View more View less
          Entrez Gene ID: (Human) 3761, (Mouse) 16520, (Rat) 116649
          View more View less
          Function(s)
          inward rectifier potassium channel activity protein binding PDZ domain binding voltage-gated ion channel activity ion channel transporter
          Process(es)
          ion transport potassium ion transport ion transmembrane transport regulation of ion transmembrane transport potassium ion import across plasma membrane transport potassium ion import cellular response to mechanical stimulus potassium ion transmembrane transport
          Pedidos Plus Icon Minus Icon
          • Estatus del pedido
          • Ayuda para pedidos
          • Orden Rápida
          • Supply Center
          • eProcurement
          Soporte Plus Icon Minus Icon
          • Ayuda y soporte
          • Entre en Contacto
          • Centros de asistencia técnica
          • Consultar documentos y certificados
          • Informar de un problema en la web
          Recursos Plus Icon Minus Icon
          • Centros de aprendizaje
          • Promociones
          • Eventos & Webinars
          • Medios Sociales
          Acerca de Thermo Fisher Plus Icon Minus Icon
          • Acerca de nosotros Acerca de nosotros
          • Empleo Empleo
          • Inversores Inversores
          • Noticias Noticias
          • Responsabilidad social Responsabilidad social
          • Marcas comerciales
          • Políticas y avisos
          Nuestro Portafolio Plus Icon Minus Icon
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline