    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Ataxin-1 (WT) Antibodies
          Antibodies Inc
          Ataxin-1/WT Monoclonal Antibody (N65/37), FL594
          View all (5) Ataxin-1 (WT) antibodies
          Tech Support
          Cite Ataxin-1/WT Monoclonal Antibody (N65/37), FL594
          Additional Information:
          {{banner.description}}
          Ataxin-1/WT Monoclonal Antibody (N65/37), FL594
          Product Details
          75-102-FL594
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N65/37
          Immunogen
          Synthetic peptide amino acids 746-761 of mouse ataxin-1 (accession number P54254).
          View immunogen
          Conjugate
          FL594 FL594 FL594
          • Unconjugated
          • FL490
          • FL550
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          594/615 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Sequence of this protein is as follows: RKRRRWSAPE TRKLEK
          Target Information
          Ataxin-1 (ATXN1) is a protein encoded by the ATXN1 gene. It is primarily known for its association with spinocerebellar ataxia type 1 (SCA1), a neurodegenerative disorder. The "WT" designation refers to the wild-type, or normal, form of the protein as opposed to mutant forms associated with disease. Ataxin-1 plays a critical role in maintaining normal neuronal function, and its dysregulation due to genetic mutations leads to severe neurodegenerative conditions. Research into Ataxin-1 continues to be vital for developing therapeutic approaches for these disorders.
          Bioinformatics
          Protein Aliases: Alt-ATXN1; alternative ataxin1; Ataxin-1; spinocerebellar ataxia 1; spinocerebellar ataxia 1 homolog; Spinocerebellar ataxia type 1 protein; Spinocerebellar ataxia type 1 protein homolog
          View more View less
          Gene Aliases: 2900016G23Rik; ATX1; ATXN1; C85907; D6S504E; ENSMUSG00000074917; Gm10786; SCA1
          View more View less
          UniProt ID: (Human) P54253, (Mouse) P54254, (Rat) Q63540
          View more View less
          Entrez Gene ID: (Human) 6310, (Mouse) 20238, (Rat) 25049
          View more View less
          Function(s)
          DNA binding chromatin binding RNA binding protein binding poly(U) RNA binding POZ domain binding poly(G) binding identical protein binding protein C-terminus binding protein self-association molecular_function
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter transcription, DNA-templated regulation of transcription, DNA-templated adult locomotory behavior visual learning negative regulation of phosphorylation negative regulation of insulin-like growth factor receptor signaling pathway negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter lung alveolus development nuclear export excitatory postsynaptic potential positive regulation of glial cell proliferation biological_process transcription from RNA polymerase II promoter RNA processing brain development learning memory social behavior insulin-like growth factor receptor signaling pathway
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline