    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CNGA1/CNGA3 Antibodies
          Antibodies Inc
          CNGA1/CNGA3 Monoclonal Antibody (L36/12), FL650
          View all (6) CNGA1/CNGA3 antibodies
          Tech Support
          Cite CNGA1/CNGA3 Monoclonal Antibody (L36/12), FL650
          Additional Information:
          {{banner.description}}
          Product Details
          75-107-FL650
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Fish, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          L36/12
          Immunogen
          Fusion protein amino acids 535-637 (cytoplasmic C-terminus) of goldfish CNG (accession number AAO16601) produced recombinantly in E. Coli.
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          CNGA1 and CNGA3 are genes that encode subunits of cyclic nucleotide-gated (CNG) channels, which are essential for sensory signal transduction in the retina. CNGA1 is primarily expressed in rod photoreceptors and is crucial for vision in low-light conditions, while CNGA3 is expressed in cone photoreceptors and is important for color vision and visual acuity. Mutations in these genes can lead to inherited visual disorders, such as retinitis pigmentosa (for CNGA1) and achromatopsia (for CNGA3).
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline