    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TRPC7 Antibodies
          Antibodies Inc
          TrpC7 Monoclonal Antibody (N64A/36), FL650
          View all (17) TRPC7 antibodies
          Tech Support
          Cite TrpC7 Monoclonal Antibody (N64A/36), FL650
          Additional Information:
          {{banner.description}}
          TrpC7 Monoclonal Antibody (N64A/36), FL650
          Product Details
          75-123-FL650
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N64A/36
          Immunogen
          Synthetic peptide amino acids 845-862 of human TrpC7 (accession number Q9HCX4).
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Sequence of this protein is as follows: FGKNLNKDHL RVNKGKDI
          Target Information
          TRPC7 is thought to form a receptor-activated non-selective calcium permeant cation channel. TRPC7 is probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. TRPC7 is also activated by diacylglycerol (DAG). Further, TRPC7 may also be activated by intracellular calcium store depletion.
          Bioinformatics
          Protein Aliases: capacitative calcium channel TRPC7; likley ortholog of mouse transient receptor potential cation channel, subfamily C, member 7; mTRP7; putative capacitative calcium channel; Short transient receptor potential channel 7; Transient receptor protein 7; TRP-7; TRP7 protein; TRPC7
          View more View less
          Gene Aliases: TRP7; TRPC7; Trrp8
          View more View less
          UniProt ID: (Human) Q9HCX4, (Mouse) Q9WVC5
          View more View less
          Entrez Gene ID: (Human) 57113, (Rat) 282822, (Mouse) 26946
          View more View less
          Function(s)
          ion channel activity calcium channel activity protein binding store-operated calcium channel activity inositol 1,4,5 trisphosphate binding cation channel activity ion channel transporter
          Process(es)
          ion transport calcium ion transport manganese ion transport single fertilization ion transmembrane transport regulation of cytosolic calcium ion concentration transmembrane transport calcium ion transmembrane transport
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline