    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • HSPA9 Antibodies
          Antibodies Inc
          Mortalin/GRP75 Monoclonal Antibody (N52A/42)
          View all (26) HSPA9 antibodies
          Tech Support
          Cite Mortalin/GRP75 Monoclonal Antibody (N52A/42)
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          Mortalin/GRP75 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          Mortalin/GRP75 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          Mortalin/GRP75 Antibody (75-127) in IHC (F)
          Adult rat hippocampus immunohistochemistry. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Mortalin/GRP75 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Mortalin/GRP75 Antibody in Western Blot (WB)
          Mortalin/GRP75 Antibody in Western Blot (WB)
          Mortalin/GRP75 Antibody in Western Blot (WB)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          75-127
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          1:200
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Non-human primate, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N52A/42
          Immunogen
          Identified as off-target mAb in screen for anti-Salm2 NeuroMabs (immunogen was fusion protein amino acids 551-766 of mouse SALM2). Target identified as Mortalin/Grp75 by LC-MS/MS analysis of protein immunopurified from rat brain using N52A/42.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4
          Contains
          0.065% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          The HSP70 family is composed of four highly conserved proteins: HSP70, HSC70, GRP75 and GRP78. These proteins serve a variety of roles. GRP75 expression is restricted to the mitochondrial matrix and aids in the translocation and folding of nascent polypeptide chains of both nuclear and mitochondrial origin. GRP75 and GRP78 are unresponsive to heat stress and are induced by glucose deprivation.
          Bioinformatics
          Protein Aliases: 70 kda heat shock protein precursor; 75 kDa glucose-regulated protein; C3H-specific antigen; catecholamine regulated protein 40; catecholamine-regulated protein 40; CRP40; epididymis secretory sperm binding protein Li 124m; GRP 75; GRP-75; heat shock 70 kDa protein 9; heat shock 70kD protein 9B; heat shock 70kDa protein 9 (mortalin); heat shock 70kDa protein 9A; Heat shock protein; heat shock protein 9; heat shock protein 9A; heat shock protein cognate 74; heat shock protein family A member 9; heat shock protein, 74 kDa, A; heat shock protein, A; HSP; MGC4500; mortalin; mortalin, perinuclear; mortalin-2; mortalin2; mtHSP70; p66 MOT; p66-mortalin; peptide-binding protein 74; stress-70 protein, mitochondrial-like; unnamed protein product
          View more View less
          Gene Aliases: 74kDa; CRP40; CSA; EVPLS; GRP-75; GRP75; HEL-S-124m; Hsc74; Hsp74; Hsp74a; Hspa9a; HSPA9B; Mortalin; MOT; Mot-2; MOT2; Mthsp70; MTHSP75; PBP74; SAAN; SIDBA4
          View more View less
          UniProt ID: (Human) P38646, (Mouse) P38647, (Rat) P48721
          View more View less
          Entrez Gene ID: (Human) 3313, (Mouse) 15526, (Rat) 291671
          View more View less
          Function(s)
          RNA binding protein binding ATP binding ATPase activity fibroblast growth factor binding enzyme binding heat shock protein binding ubiquitin protein ligase binding protein binding involved in protein folding chaperone binding
          Process(es)
          protein folding protein export from nucleus response to toxic substance iron-sulfur cluster assembly erythrocyte differentiation calcium ion transmembrane import into mitochondrion protein refolding positive regulation of apoptotic process regulation of erythrocyte differentiation negative regulation of erythrocyte differentiation protein autophosphorylation response to folic acid cellular response to interleukin-1 response to thyroxine negative regulation of hematopoietic stem cell differentiation negative regulation of hemopoiesis intracellular protein transport inner mitochondrial membrane organization protein import into mitochondrial matrix negative regulation of apoptotic process
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-74676f44b6-xtlnb:80/100.66.72.150:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0