    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • FGF11 Antibodies
          Antibodies Inc
          FGF11/FHF3 Monoclonal Antibody (N92/14), FL650
          View all (11) FGF11 antibodies
          Tech Support
          Cite FGF11/FHF3 Monoclonal Antibody (N92/14), FL650
          Additional Information:
          {{banner.description}}
          FGF11/FHF3 Monoclonal Antibody (N92/14), FL650
          Product Details
          75-133-FL650
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N92/14
          Immunogen
          Fusion protein amino acids 55-216 of human FGF11a (accession number Q92914) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Probably involved in nervous system development and function.
          Bioinformatics
          Protein Aliases: FGF; FGF-11; FHF-3; Fibroblast growth factor; Fibroblast growth factor 11; Fibroblast growth factor homologous factor 3; unnamed protein product
          View more View less
          Gene Aliases: FGF-11; FGF11; FHF-3; FHF3
          View more View less
          UniProt ID: (Human) Q92914, (Mouse) P70378
          View more View less
          Entrez Gene ID: (Human) 2256, (Mouse) 14166
          View more View less
          Function(s)
          protein binding growth factor activity sodium channel regulator activity receptor binding growth factor intercellular signal molecule
          Process(es)
          signal transduction cell-cell signaling nervous system development neurogenesis
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline