    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TMEM91 Antibodies
          Antibodies Inc
          SynDIG3/Tmem91 Monoclonal Antibody (N160/21), FL490
          View all (8) TMEM91 antibodies
          Tech Support
          Cite SynDIG3/Tmem91 Monoclonal Antibody (N160/21), FL490
          Additional Information:
          {{banner.description}}
          SynDIG3/Tmem91 Monoclonal Antibody (N160/21), FL490
          Product Details
          75-186-FL490
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N160/21
          Immunogen
          Fusion protein amino acids 1-96 (cytoplasmic N-terminus) of mouse SynDIG3 (accession number Q8C581) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL490 FL490 FL490
          • Unconjugated
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          491/515 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          The function remains unknown.
          Bioinformatics
          Protein Aliases: Dispanin subfamily C member 3; DSPC3; Synapse differentiation-induced protein 3; Transmembrane protein 91
          View more View less
          Gene Aliases: A830041P22Rik; DSPC3; SynDIG3; Tmem91
          View more View less
          UniProt ID: (Mouse) Q8C581
          View more View less
          Entrez Gene ID: (Mouse) 320208, (Rat) 100359879
          View more View less
          Function(s)
          molecular_function
          Process(es)
          hematopoietic progenitor cell differentiation response to biotic stimulus
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline