    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • S100A5 Antibodies
          Antibodies Inc
          S100A5 Monoclonal Antibody (N176/35), FL650
          View all (23) S100A5 antibodies
          Tech Support
          Cite S100A5 Monoclonal Antibody (N176/35), FL650
          Additional Information:
          {{banner.description}}
          S100A5 Monoclonal Antibody (N176/35), FL650
          Product Details
          75-199-FL650
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N176/35
          Immunogen
          Fusion protein amino acids 1-93 (full-length) of rat S100A (accession number P63083) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Binds calcium, zinc and copper. One subunit can simultaneously bind 2 calcium ions or 2 copper ions plus 1 zinc ion. Calcium and copper ions compete for the same binding sites. Engelkamp D., Proc. Natl. Acad. Sci. U.S.A. 90:6547-6551(1993). Gregory S.G., Nature 441:315-321(2006).
          Bioinformatics
          Protein Aliases: Protein S-100D; Protein S100-A5; S100 calcium-binding protein A5
          View more View less
          Gene Aliases: S100a5; S100d; S100D9
          View more View less
          UniProt ID: (Rat) P63083, (Mouse) P63084
          View more View less
          Entrez Gene ID: (Rat) 295211, (Mouse) 20199
          View more View less
          Function(s)
          copper ion binding calcium ion binding zinc ion binding protein homodimerization activity metal ion binding calmodulin-related
          Process(es)
          biological_process
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline