    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • Neurexin-1-beta Antibodies
          Antibodies Inc
          Neurexin-1-Beta Monoclonal Antibody (N170A/1), FL550
          View all (8) Neurexin-1-beta antibodies
          Tech Support
          Cite Neurexin-1-Beta Monoclonal Antibody (N170A/1), FL550
          Additional Information:
          {{banner.description}}
          Product Details
          75-216-FL550
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N170A/1
          Immunogen
          Synthetic peptide amino acids 47-64 of rat Neurexin-1-Beta (accession number Q63373).
          Conjugate
          FL550 FL550 FL550
          • Unconjugated
          • FL490
          • FL594
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          550/575 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Sequence of this protein is as follows: ASSLGAHHIH HFHGSSKH
          Target Information
          Neurexin-1-beta (NRXN1-beta) is a gene that encodes the beta-isoform of the neurexin-1 protein, which is a type I membrane protein that functions as a cell adhesion molecule and receptor in the nervous system. Located on chromosome 2 in humans, the neurexin-1 gene undergoes complex alternative splicing, producing multiple isoforms, including the well-studied alpha-neurexin and beta-neurexin. Neurexins, including NRXN1-beta, are primarily involved in synapse formation and function by mediating interactions between presynaptic and postsynaptic neurons. They bind to neuroligins, another family of cell adhesion molecules, thus playing a critical role in the maintenance and regulation of synaptic specificity and neurotransmitter release. Dysregulation or mutations in NRXN1-beta have been implicated in various neurological and psychiatric disorders, including autism spectrum disorders, schizophrenia, and intellectual disabilities, emphasizing the importance of this gene in brain development and function.
          Bioinformatics
          Protein Aliases: alpha-latrotoxin receptor (calcium-dependent); neurexin 1; neurexin I; Neurexin I-alpha; Neurexin I-beta; Neurexin-1; Neurexin-1-alpha; Neurexin-1-beta; neuronal cell surface protein; non-processed neurexin I-alpha; unnamed protein product
          View more View less
          Gene Aliases: 1700062G21Rik; 9330127H16Rik; A230068P09Rik; Hs.22998; KIAA0578; mKIAA0578; NRXN1; Nrxn1b; PTHSL2; SCZD17
          View more View less
          UniProt ID: (Human) Q9ULB1, (Mouse) Q9CS84, (Rat) Q63372
          View more View less
          Entrez Gene ID: (Human) 9378, (Mouse) 18189, (Rat) 60391
          View more View less
          Function(s)
          transmembrane signaling receptor activity receptor binding type 1 fibroblast growth factor receptor binding calcium channel regulator activity calcium ion binding protein binding acetylcholine receptor binding signaling receptor activity metal ion binding calcium-dependent protein binding cell adhesion molecule binding neuroligin family protein binding
          Process(es)
          angiogenesis cell adhesion heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules neuron cell-cell adhesion signal transduction chemical synaptic transmission neurotransmitter secretion axon guidance synapse assembly learning positive regulation of gene expression negative regulation of gene expression calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules cerebellar granule cell differentiation neuronal signal transduction adult behavior neuron projection development positive regulation of synaptic transmission, GABAergic social behavior protein localization to synapse vocal learning positive regulation of cAMP-mediated signaling establishment of protein localization positive regulation of fibroblast growth factor receptor signaling pathway neuron projection morphogenesis neuromuscular process controlling balance negative regulation of filopodium assembly positive regulation of protein kinase B signaling positive regulation of synapse assembly positive regulation of synaptic transmission, glutamatergic positive regulation of ERK1 and ERK2 cascade cellular response to calcium ion vocalization behavior protein complex assembly involved in synapse maturation positive regulation of synapse maturation synaptic vesicle clustering postsynaptic membrane assembly presynaptic membrane assembly gamma-aminobutyric acid receptor clustering NMDA glutamate receptor clustering gephyrin clustering involved in postsynaptic density assembly guanylate kinase-associated protein clustering neuroligin clustering involved in postsynaptic membrane assembly postsynaptic density protein 95 clustering receptor localization to synapse NMDA selective glutamate receptor signaling pathway AMPA selective glutamate receptor signaling pathway positive regulation of neuromuscular synaptic transmission positive regulation of phospholipase C-activating G-protein coupled receptor signaling pathway positive regulation of protein localization to plasma membrane positive regulation of presynaptic active zone assembly positive regulation of excitatory postsynaptic potential vesicle docking involved in exocytosis protein heterotetramerization prepulse inhibition regulation of insulin secretion involved in cellular response to glucose stimulus regulation of grooming behavior positive regulation of establishment of protein localization to plasma membrane regulation of N-methyl-D-aspartate selective glutamate receptor activity regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline