          • Primary Antibodies ›
          • Kirrel3 Short Isoform 3 Antibodies
          Antibodies Inc
          Kirrel3 Short Isoform 3 Monoclonal Antibody (N320/48), FL490
          View all (5) Kirrel3 Short Isoform 3 antibodies
          Tech Support
          Cite Kirrel3 Short Isoform 3 Monoclonal Antibody (N320/48), FL490
          Additional Information:
          {{banner.description}}
          Kirrel3 Short Isoform 3 Monoclonal Antibody (N320/48), FL490
          Product Details
          75-282-FL490
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N320/48
          Immunogen
          Synthetic peptide amino acids 591-608 of mouse Kirrel3 short isoform 3 (accession number Q8BR86-3).
          3D Epitope / Immunogen
          Conjugate
          FL490 FL490 FL490
          • Unconjugated
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          491/515 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Sequence of this protein is as follows: VRAQPMPHSI LSTQTSRC
          Target Information
          Kirrel3 Short Isoform 3 is a variant of the Kirrel3 protein, which is a member of the immunoglobulin superfamily involved in cell adhesion and signaling. This specific isoform, as a shorter version, may have distinct functional or regulatory roles compared to the full-length protein, particularly in the development and maintenance of neural connections. Mutations or dysregulation of Kirrel3 have been associated with neurodevelopmental disorders, including intellectual disability and autism spectrum disorders.
          Bioinformatics
          Protein Aliases: Kin of IRRE-like protein 3; Kin of irregular chiasm-like protein 3; membrane protein mKirre; mKirre; Nephrin-like protein 2; X kin of IRRE like 3
          View more View less
          Gene Aliases: 1500010O20Rik; 2900036G11Rik; Kiaa1867; Kirrel3; mKIAA1867; NEPH2; SST4
          View more View less
          UniProt ID: (Mouse) Q8BR86
          View more View less
          Entrez Gene ID: (Mouse) 67703
          View more View less
          Function(s)
          protein binding PDZ domain binding
          Process(es)
          neuron migration inter-male aggressive behavior principal sensory nucleus of trigeminal nerve development hemopoiesis neuron projection morphogenesis glomerulus morphogenesis
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline