    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Synaptotagmin 9 Antibodies
          Antibodies Inc
          Synaptotagmin-9 Monoclonal Antibody (N276A/15), FL650
          View all (13) Synaptotagmin 9 antibodies
          Tech Support
          Cite Synaptotagmin-9 Monoclonal Antibody (N276A/15), FL650
          Additional Information:
          {{banner.description}}
          Synaptotagmin-9 Monoclonal Antibody (N276A/15), FL650
          Product Details
          75-306-FL650
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N276A/15
          Immunogen
          Fusion protein amino acids 253-344 (cytoplasmic C2B domain) of mouse Synaptotagmin-9 (accession number Q9R0N5) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          SYT9 may be involved in Ca2+-dependent exocytosis of secretory vesicles through Ca2+ and phospholipid binding to the C2 domain or may serve as Ca2+ sensors in the process of vesicular trafficking and exocytosis.
          Bioinformatics
          Protein Aliases: FLJ45896; synaptogamin V; Synaptotagmin 5; synaptotagmin 9; Synaptotagmin IX; Synaptotagmin V; Synaptotagmin-9; Synaptotagmin9; SynaptotagminIX; Syt IX; SYT-9; SytIX; unnamed protein product
          View more View less
          Gene Aliases: Syt5; SYT9; Sytv
          View more View less
          UniProt ID: (Human) Q86SS6, (Mouse) Q9R0N9, (Rat) Q925C0
          View more View less
          Entrez Gene ID: (Human) 143425, (Mouse) 60510, (Rat) 60564
          View more View less
          Function(s)
          SNARE binding phosphatidylserine binding protein binding calcium-dependent phospholipid binding phosphatidylinositol-4,5-bisphosphate binding identical protein binding metal ion binding calcium ion sensor activity calcium ion binding syntaxin binding clathrin binding transporter activity membrane trafficking regulatory protein
          Process(es)
          chemical synaptic transmission vesicle-mediated transport regulation of calcium ion-dependent exocytosis positive regulation of vesicle fusion calcium-dependent activation of synaptic vesicle fusion synaptic vesicle exocytosis positive regulation of calcium ion-dependent exocytosis calcium ion-regulated exocytosis of neurotransmitter regulation of insulin secretion detection of calcium ion transport
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline