    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • BAF53B Antibodies
          Antibodies Inc
          BAF53b Monoclonal Antibody (N332B/15)
          View all (11) BAF53B antibodies
          Tech Support
          Cite BAF53b Monoclonal Antibody (N332B/15)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          BAF53b Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          BAF53b Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          BAF53b Antibody (75-311) in WB
          Tissue and transfected cell immunoblot: extracts of rat brain low speed pellet (RBLSP) and COS cells transiently transfected with untagged BAF53a, BAF53b or Kv2.1 plasmids and probed with N332B/15 (left) and N52A/42 (right) TC supe. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          BAF53b Antibody in Western Blot (WB)
          BAF53b Antibody in Western Blot (WB)
          BAF53b Monoclonal Antibody (N332B/15)
          Product Details
          75-311
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          1:500
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N332B/15
          Immunogen
          Fusion protein amino acids 39-114 of human BAF53b (accession number O94805) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4
          Contains
          0.065% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Sequence of this protein is as follows: TTVGLLAAEE GGGLELEGDK EKKGKIFHID TNALHVPRDG AEVMSPLKNG MIEDWECFRA ILDHTYSKHV KSEPNL
          Target Information
          Belongs to the chromatin remodeling brain-specific BAF (bBAF) complex, as such plays a role in remodeling mononucleosomes in an ATP-dependent fashion. Belongs to the neuron-specific chromatin remodeling complex (nBAF complex) and is required for postmitotic neural development and dendritic outgrowth. During neural development a switch from a stem/progenitor to a post-mitotic chromatin remodeling mechanism occurs as neurons exit the cell cycle and become committed to their adult state. The transition from proliferating neural stem/progenitor cells to post-mitotic neurons requires a switch in subunit composition of the npBAF and nBAF complexes. As neural progenitors exit mitosis and differentiate into neurons, npBAF complexes which contain ACTL6A/BAF53A and PHF10/BAF45A, are exchanged for homologous alternative ACTL6B/BAF53B and DPF1/BAF45B or DPF3/BAF45C subunits in neuron-specific complexes (nBAF). The npBAF complex is essential for the self-renewal/proliferative capacity of the multipotent neural stem cells. The nBAF complex along with CREST plays a role regulating the activity of genes essential for dendrite growth. ACTL6B/BAF53B is not essential for assembly of the nBAF complex but is required for targeting the complex and CREST to the promoter of genes essential for dendritic growth.
          Bioinformatics
          Protein Aliases: 53 kDa BRG1-associated factor B; actin-like 6; Actin-like protein 6B; Actin-related protein Baf53b; ArpN-alpha; ArpNa; ArpNalpha; BAF53B; BRG1-associated factor 53B; hArpN alpha; unnamed protein product
          View more View less
          Gene Aliases: ACTL6; ACTL6B; ArpNa; arpNalpha; BAF53B; DEE76; EIEE76; IDDSSAD; SMARCN2
          View more View less
          UniProt ID: (Human) O94805, (Mouse) Q99MR0
          View more View less
          Entrez Gene ID: (Human) 51412, (Mouse) 83766
          View more View less
          Function(s)
          chromatin binding transcription coactivator activity structural constituent of cytoskeleton protein binding actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein
          Process(es)
          chromatin organization chromatin remodeling regulation of transcription from RNA polymerase II promoter cytoskeleton organization nervous system development positive regulation of cell proliferation dendrite development regulation of mitotic metaphase/anaphase transition neuron maturation positive regulation of T cell differentiation negative regulation of cell differentiation positive regulation of cell differentiation positive regulation of myoblast differentiation positive regulation of transcription, DNA-templated regulation of G0 to G1 transition positive regulation of stem cell population maintenance regulation of G1/S transition of mitotic cell cycle positive regulation of double-strand break repair regulation of nucleotide-excision repair chromatin assembly or disassembly transcription, DNA-templated regulation of transcription, DNA-templated chromatin modification
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline