    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • PSD Antibodies
          Antibodies Inc
          PSD Marker Monoclonal Antibody (N286/74), FL490
          View all (10) PSD antibodies
          Tech Support
          Cite PSD Marker Monoclonal Antibody (N286/74), FL490
          Additional Information:
          {{banner.description}}
          Product Details
          75-312-FL490
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Zebrafish
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N286/74
          Immunogen
          Heterogeneous postsynaptic density fraction of adult zebrafish brain. Specific protein target(s) not yet identified but likely to be a MAGUK protein.
          Conjugate
          FL490 FL490 FL490
          • Unconjugated
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          491/515 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          PSD, which called EFA6A, is 63% identical to EFA6B (PSD4) and contains a Sec7 domain adjacent to a PH domain, followed by a C-terminal coiled-coil motif. This gene is mapped to chromosome 10q24. And it encodes a Plekstrin homology and SEC7 domains-containing protein that functions as a guanine nucleotide exchange factor. The encoded protein regulates signal transduction by activating ADP-ribosylation factor 6. Alternative splicing results in multiple transcript variants.
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline