    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • KIRREL3 Antibodies
          Antibodies Inc
          Kirrel3 Short/Long Monoclonal Antibody (N321C/49)
          View all (11) KIRREL3 antibodies
          Tech Support
          Cite Kirrel3 Short/Long Monoclonal Antibody (N321C/49)
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          Kirrel3 Short/Long Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Kirrel3 Short/Long Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Kirrel3 Short/Long Antibody (75-333) in WB
          Tissue and transfected cell immunoblot: extracts of rat brain membrane (RBM) and COS cells transiently transfected with Flag-tagged Kirrel3-short, Kirrel3-long, untagged Kirrel2, Kirrel1 or Kv2.1 plasmid and probed with N321C/49 (left) and N52A/42 (right) TC supe. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Kirrel3 Short/Long Antibody in Western Blot (WB)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          75-333
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N321C/49
          Immunogen
          Fusion protein amino acids 46-524 (extracellular N-terminus) of rat Kirrel3 (accession number Q09GS6) produced recombinantly in E. Coli.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4
          Contains
          0.065% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          The protein encoded by this gene is a member of the nephrin-like protein family. These proteins are expressed in fetal and adult brain, and also in podocytes of kidney glomeruli. The cytoplasmic domains of these proteins interact with the C-terminus of podocin, also expressed in the podocytes, cells involved in ensuring size- and charge-selective ultrafiltration. The protein encoded by this gene is a synaptic cell adhesion molecule with multiple extracellular immunoglobulin-like domains and a cytoplasmic PDZ domain-binding motif. Mutations in this gene are associated with several neurological and cognitive disorders. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
          Bioinformatics
          Protein Aliases: kin of IRRE like 3; kin of irregular chiasm-like protein 3; membrane protein mKirre; nephrin-like protein 2; rKirre; SST4; unnamed protein product; X kin of IRRE like 3
          View more View less
          Gene Aliases: 1500010O20Rik; 2900036G11Rik; mKIAA1867; NEPH2; SST4
          View more View less
          UniProt ID: (Mouse) Q8BR86
          View more View less
          Entrez Gene ID: (Rat) 315546, (Mouse) 67703
          View more View less
          Function(s)
          cell adhesion molecule binding protein binding PDZ domain binding
          Process(es)
          neuron migration inter-male aggressive behavior homophilic cell adhesion via plasma membrane adhesion molecules synapse assembly principal sensory nucleus of trigeminal nerve development hippocampus development hemopoiesis neuron projection morphogenesis glomerulus morphogenesis cell-cell adhesion
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-7bf6cff7cf-xbl4w:80/100.66.75.228:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0