    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • STON2 Antibodies
          Antibodies Inc
          Stonin-2 Monoclonal Antibody (N346/9)
          View all (6) STON2 antibodies
          Tech Support
          Cite Stonin-2 Monoclonal Antibody (N346/9)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Stonin-2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Stonin-2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Stonin-2 Antibody (75-337) in WB
          Transfected cell immunoblot: extracts of COS cells transiently transfected with HA-tagged Stonin-1, Stonin-2 or untagged Kv2.1 plasmid and probed with N346/9 TC supe (left), Rabbit anti-HA (middle) or N52A/42 TC supe (right). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Stonin-2 Antibody in Western Blot (WB)
          Stonin-2 Monoclonal Antibody (N346/9)
          Product Details
          75-337
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N346/9
          Immunogen
          Fusion protein amino acids 1-555 (N-terminus) of human Stonin-2 (accession number Q8WXE9) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4
          Contains
          0.065% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Bioinformatics
          Protein Aliases: homolog of stoned B; similar to stoned B; Stoned B; stoned B homolog 2; Stonin-2
          View more View less
          Gene Aliases: 4933401N24Rik; AI425999; AW544599; RGD1311502; STN2; STNB; STNB2; STON2
          View more View less
          UniProt ID: (Human) Q8WXE9, (Mouse) Q8BZ60, (Rat) D4AB66
          View more View less
          Entrez Gene ID: (Human) 85439, (Mouse) 108800, (Rat) 314349
          View more View less
          Function(s)
          protein binding clathrin adaptor activity membrane traffic protein
          Process(es)
          endocytosis regulation of endocytosis synaptic vesicle endocytosis clathrin-mediated endocytosis hematopoietic progenitor cell differentiation intracellular protein transport chemical synaptic transmission vesicle-mediated transport
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0