    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SPTB Antibodies
          Antibodies Inc
          Beta1-Spectrin Monoclonal Antibody (N385/21), FL650
          View all (12) SPTB antibodies
          Tech Support
          Cite Beta1-Spectrin Monoclonal Antibody (N385/21), FL650
          Additional Information:
          {{banner.description}}
          Beta1-Spectrin Monoclonal Antibody (N385/21), FL650
          Product Details
          75-374-FL650
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N385/21
          Immunogen
          Fusion protein amino acids 1555-2137 (C-terminal repeats 13 to 17) of human Beta1-spectrin (accession number P11277) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          SPTB (Spectrin Beta, Erythrocytic) is a Protein Coding gene. Diseases associated with SPTB include Spherocytosis, Type 2 and Elliptocytosis 3. Among its related pathways are Cytokine Signaling in Immune system and RET signaling. Gene Ontology (GO) annotations related to this gene include protein heterodimerization activity and actin filament binding. An important paralog of this gene is SPTBN1.
          Bioinformatics
          Protein Aliases: beta-chain variant; Beta-I spectrin; beta-spectrin; beta-spectrin 1; brain erythroid spectrin (235E); erythrocyte beta-spectrin; erythrocyte membrane protein; erythrocytic spectrin beta chain; erythroid spectrin beta; HSPTB1; membrane cytoskeletal protein; Sp beta; spectrin beta 1; spectrin beta chain, erythrocyte; Spectrin beta chain, erythrocytic; Spectrin Beta Erythrocytic; spectrin beta Tandil; Spectrin Beta, Erythrocytic; Spectrin is the major constituent of the cytoskeletal network underlying the erythrocyte plasma membrane.; spectrin R; spectrin Rouen (beta-220-218) mutant coding sequence; SPTB1; unnamed protein product
          View more View less
          Gene Aliases: AI842465; D330027P03Rik; EL3; Gm1301; HS2; HSPTB1; ja; jaundiced; mKIAA4219; SPH2; Spnb-1; Spnb1; SPTB; SPTB1
          View more View less
          UniProt ID: (Human) P11277
          View more View less
          Entrez Gene ID: (Human) 6710, (Rat) 314251, (Mouse) 20741
          View more View less
          Function(s)
          actin binding structural constituent of cytoskeleton protein binding ankyrin binding actin filament binding molecular_function protein heterodimerization activity
          Process(es)
          porphyrin-containing compound biosynthetic process plasma membrane organization biological_process hemopoiesis actin filament capping actin cytoskeleton organization synapse organization modification of postsynaptic actin cytoskeleton
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline