    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • GLRA3 Antibodies
          Antibodies Inc
          Glycine Receptor Alpha3 Monoclonal Antibody (N424/45), FL650
          View all (13) GLRA3 antibodies
          Tech Support
          Cite Glycine Receptor Alpha3 Monoclonal Antibody (N424/45), FL650
          Additional Information:
          {{banner.description}}
          Glycine Receptor Alpha3 Monoclonal Antibody (N424/45), FL650
          Product Details
          75-417-FL650
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N424/45
          Immunogen
          Fusion protein amino acids 342-431 (cytoplasmic loop) of human GlyRAlpha3L (accession number O75311) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          The GLRA3 gene encodes the alpha-3 subunit of the neuronal glycine receptor, a ligand-gated chloride channel composed of ligand-binding alpha and structural beta polypeptides.
          Bioinformatics
          Protein Aliases: glycine receptor; Glycine receptor subunit alpha-3; glycine receptor, alpha 3 subunit; glycine receptor, alpha-3 polypeptide; ligand gated ion channel
          View more View less
          Gene Aliases: GLRA3
          View more View less
          UniProt ID: (Human) O75311, (Mouse) Q91XP5
          View more View less
          Entrez Gene ID: (Human) 8001, (Rat) 114516, (Mouse) 110304
          View more View less
          Function(s)
          transmembrane signaling receptor activity ion channel activity extracellular ligand-gated ion channel activity excitatory extracellular ligand-gated ion channel activity chloride channel activity protein binding glycine binding extracellular-glycine-gated chloride channel activity transmitter-gated ion channel activity glycine-gated chloride ion channel activity metal ion binding ligand-gated ion channel
          Process(es)
          ion transport chloride transport ion transmembrane transport protein homooligomerization excitatory postsynaptic potential presynaptic modulation of chemical synaptic transmission chloride transmembrane transport receptor clustering neurotransmitter-gated ion channel clustering gamma-aminobutyric acid receptor clustering transport
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline