    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
          • Primary Antibodies ›
          • KCNV2 Antibodies
          Antibodies Inc
          Kv8.2 Potassium Channel Monoclonal Antibody (N448/88), FL594
          View all (12) KCNV2 antibodies
          Tech Support
          Cite Kv8.2 Potassium Channel Monoclonal Antibody (N448/88), FL594
          Additional Information:
          {{banner.description}}
          Kv8.2 Potassium Channel Monoclonal Antibody (N448/88), FL594
          Product Details
          75-435-FL594
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N448/88
          Immunogen
          Fusion protein amino acids 1-163 (cytoplasmic N-terminus) of mouse Kv8.2 (accession number Q8CFS6) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL594 FL594 FL594
          • Unconjugated
          • FL490
          • FL550
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          594/615 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          KCNV2 is a potassium channel subunit. Modulates channel activity by shifting the threshold and the half-maximal activation to more negative values.
          Bioinformatics
          Protein Aliases: Potassium voltage-gated channel subfamily V member 2; Voltage-gated potassium channel subunit Kv8.2
          View more View less
          Gene Aliases: Kcnv2; KV11.1
          View more View less
          UniProt ID: (Mouse) Q8CFS6
          View more View less
          Entrez Gene ID: (Mouse) 240595
          View more View less
          Function(s)
          ion channel activity voltage-gated ion channel activity voltage-gated potassium channel activity delayed rectifier potassium channel activity potassium channel activity voltage-gated ion channel
          Process(es)
          transport ion transport potassium ion transport regulation of ion transmembrane transport protein homooligomerization transmembrane transport potassium ion transmembrane transport
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline