    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
          • Primary Antibodies ›
          • ICK Antibodies
          Antibodies Inc
          ICK Monoclonal Antibody (N451/73), FL490
          View all (15) ICK antibodies
          Tech Support
          Cite ICK Monoclonal Antibody (N451/73), FL490
          Additional Information:
          {{banner.description}}
          ICK Monoclonal Antibody (N451/73), FL490
          Product Details
          75-437-FL490
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N451/73
          Immunogen
          Fusion protein amino acids 290-629 (C-terminus) of mouse ICK (accession number Q9JKV2) produced recombinantly in E. Coli.
          View immunogen
          Conjugate
          FL490 FL490 FL490
          • Unconjugated
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          491/515 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          ICK or intestinal cell kinase belongs to a very extensive family of proteins that share a conserved catalytic core common with both serine/threonine and tyrosine protein kinases. ICK encodes an intestinal serine/threonine kinase harboring a dual phosphorylation site found in mitogen-activating protein (MAP) kinases. ICK localizes to the intestinal crypt region and is thought to be important in intestinal epithelial cell proliferation and differentiation. ICK purified from transfected human embryonic kidney cells undergoes autophosphorylation and phosphorylates a test protein. ICK plays a key role in development of central nervous, skeletal, and endocrine systems.
          Bioinformatics
          Protein Aliases: heart serine-threonine protein kinase; Intestinal cell kinase; MAK-related kinase; mICK; MRK; Serine/threonine-protein kinase ICK
          View more View less
          Gene Aliases: 2210420N10Rik; AI848300; Cilk1; Ick; Mrk
          View more View less
          UniProt ID: (Rat) Q62726, (Mouse) Q9JKV2
          View more View less
          Entrez Gene ID: (Rat) 84411, (Mouse) 56542
          View more View less
          Function(s)
          magnesium ion binding protein serine/threonine kinase activity cyclin-dependent protein serine/threonine kinase activity ATP binding metal ion binding nucleotide binding protein kinase activity kinase activity transferase activity transferase activity, transferring phosphorus-containing groups non-receptor serine/threonine protein kinase
          Process(es)
          protein phosphorylation multicellular organism development intracellular signal transduction intraciliary transport cilium assembly regulation of cell cycle signal transduction phosphorylation cell projection organization
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0