    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • Neuroligin 2 Antibodies
          Antibodies Inc
          Neuroligin-2 Monoclonal Antibody (L107/39), FL594
          View all (13) Neuroligin 2 antibodies
          Tech Support
          Cite Neuroligin-2 Monoclonal Antibody (L107/39), FL594
          Additional Information:
          {{banner.description}}
          Neuroligin-2 Monoclonal Antibody (L107/39), FL594
          Product Details
          75-451-FL594
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          L107/39
          Immunogen
          Fusion protein amino acids 700-836 (cytoplasmic C-terminus) of mouse Neuroligin-2 (accession number Q69ZK9) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL594 FL594 FL594
          • Unconjugated
          • FL490
          • FL550
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          594/615 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses.
          Bioinformatics
          Protein Aliases: HLN2; Neuroligin-2; ortholog of human and rat neuroligin 2 NLGN2
          View more View less
          Gene Aliases: KIAA1366; NL2; NLGN2
          View more View less
          UniProt ID: (Human) Q8NFZ4, (Mouse) Q69ZK9
          View more View less
          Entrez Gene ID: (Human) 57555, (Mouse) 216856
          View more View less
          Function(s)
          neurexin family protein binding identical protein binding cell adhesion molecule binding receptor activity protein binding carboxylic ester hydrolase activity cell adhesion molecule
          Process(es)
          thigmotaxis regulation of respiratory gaseous exchange by neurological system process cell adhesion neuron cell-cell adhesion synapse assembly jump response positive regulation of cell proliferation sensory perception of pain positive regulation of insulin secretion positive regulation of synaptic transmission, GABAergic protein localization to cell surface social behavior protein localization to synapse locomotory exploration behavior cell-cell junction maintenance modulation of synaptic transmission synapse organization neuromuscular process controlling balance synaptic transmission, GABAergic positive regulation of synapse assembly positive regulation of synaptic transmission, glutamatergic positive regulation of dendritic spine development terminal button organization neurotransmitter-gated ion channel clustering postsynaptic membrane assembly presynaptic membrane assembly gephyrin clustering involved in postsynaptic density assembly postsynaptic density protein 95 clustering positive regulation of inhibitory postsynaptic potential cell-cell adhesion postsynaptic specialization assembly presynapse assembly insulin metabolic process positive regulation of protein localization to synapse positive regulation of t-SNARE clustering inhibitory synapse assembly regulation of presynapse assembly positive regulation of excitatory postsynaptic potential positive regulation of synaptic vesicle clustering regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline