    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • Parvalbumin Antibodies
          Antibodies Inc
          Parvalbumin Monoclonal Antibody (L114/3), FL650
          View all (41) Parvalbumin antibodies
          Tech Support
          Cite Parvalbumin Monoclonal Antibody (L114/3), FL650
          Additional Information:
          {{banner.description}}
          Parvalbumin Monoclonal Antibody (L114/3), FL650
          Product Details
          75-455-FL650
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          L114/3
          Immunogen
          Fusion protein amino acids 1-110 (full-length) of rat Parvalbumin (accession number P02625) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Parvalbumin (PV) is a calcium binding protein expressed in specific muscle fibers and fast-firing neurons. PV consists of a single, unbranched chain of linked amino acids and belongs to a larger group of EF hand proteins. Studies have demonstrated that parvalbumin acts in the decay of calcium in the contraction/ relaxation cycle of fast twitch muscles. This data has shown a positive correlation between the rate of relaxation and the concentration of parvalbumin. Parvalbumin is also expressed in a specific population of GABAergic interneurones which are thought to play a role in maintaining the balance between excitation and inhibition in the cortex as well as the hippocampus. In amyotrophic lateral sclerosis (ALS) patents, parvalbumin immunoreactivity is specifically absent from neuron populations lost early in ALS.
          Bioinformatics
          Protein Aliases: Alpha-parvalbumin; Alpha-PV; HGNC:9704; MGC116759; Parvalbumin (calcium binding protein); Parvalbumin alpha
          View more View less
          Gene Aliases: PALB1; Parv; PV; Pva; Pvalb
          View more View less
          UniProt ID: (Rat) P02625, (Mouse) P32848
          View more View less
          Entrez Gene ID: (Rat) 25269, (Mouse) 19293
          View more View less
          Function(s)
          calcium ion binding protein homodimerization activity protein heterodimerization activity metal ion binding calmodulin-related
          Process(es)
          regulation of cytosolic calcium ion concentration
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline