    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • KCNK6 Antibodies
          Antibodies Inc
          TWIK-2 Monoclonal Antibody (N469/28), FL594
          View all (13) KCNK6 antibodies
          Tech Support
          Cite TWIK-2 Monoclonal Antibody (N469/28), FL594
          Additional Information:
          {{banner.description}}
          TWIK-2 Monoclonal Antibody (N469/28), FL594
          Product Details
          75-463-FL594
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N469/28
          Immunogen
          Fusion protein amino acids 28-92 (first extracellular loop) of mouse TWIK-2 (accession number Q3TBV4) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL594 FL594 FL594
          • Unconjugated
          • FL490
          • FL550
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          594/615 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          KCNK6 channels play a critical role in maintaining the electrical stability of cells, and their dysfunction can contribute to a variety of pathophysiological conditions. KCNK6, also known as TWIK-2 (Tandem of Pore Domains in a Weak Inwardly Rectifying K+ Channel 2), is a member of the two-pore-domain potassium (K2P) channel family.
          Bioinformatics
          Protein Aliases: FLJ12282; Potassium channel subfamily K member 6; TWIK-2 two-pore-domain K+ channel
          View more View less
          Gene Aliases: D7Ertd764e; Kcnk6; Toss; Twik2
          View more View less
          UniProt ID: (Mouse) Q3TBV4
          View more View less
          Entrez Gene ID: (Mouse) 52150
          View more View less
          Function(s)
          potassium channel activity potassium ion leak channel activity ion channel transporter
          Process(es)
          regulation of systemic arterial blood pressure negative regulation of systemic arterial blood pressure stabilization of membrane potential regulation of resting membrane potential potassium ion transmembrane transport
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0