    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • VAPA/VAPB Antibodies
          Antibodies Inc
          VAPA/B Monoclonal Antibody (N479/107), FL650
          View all (7) VAPA/VAPB antibodies
          Tech Support
          Cite VAPA/B Monoclonal Antibody (N479/107), FL650
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          VAPA/B Monoclonal Antibody (N479/107), FL650
          Product Details
          75-496-FL650
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N479/107
          Immunogen
          Fusion protein amino acids 1-219 (N-terminus) of rat VAPA (accession number Q9Z270) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          VAPA/VAPB may play a role in vesicle trafficking. Present in the plasma membrane and in intracellular vesicles. Interacts with VAMP1 and VAMP2. Subunit: Homodimer, and heterodimer with VAPA. Subcellular Location: Cell membrane; Single-pass type IV membrane protein. Intracytoplasmic membrane; Single-pass type IV membrane protein. Tissue Specificity: Ubiquitous. Isoform 1 predominates. Disease: Defects in VAPB are the cause of amyotrophic lateral sclerosis type 8 (ALS8). ALS8 is a familial form of amyotrophic lateral sclerosis, a neurodegenerative disorder affecting upper and lower motor neurons and resulting in fatal paralysis. Sensory abnormalities are absent. Death usually occurs within 2 to 5 years. The etiology of amyotrophic lateral sclerosis is likely to be multifactorial, involving both genetic and environmental factors. The disease is inherited in 5-10% of cases leading to familial forms. Defects in VAPB are a cause of proximal adult autosomal dominant spinal muscular atrophy; also called late-onset spinal muscular atrophy Finkel type. Spinal muscular atrophies are neurodegenerative disorders characterized by degeneration of lower motor neurons, leading to progressive paralysis muscular atrophy. This form is a late- adult-onset form of the disease (after age 20 years). The patients show a benign course, most of them remaining ambulatory 10 to 40 years after clinical onset.
          Bioinformatics
          Protein Aliases: 33 kDa VAMP-associated protein; similar to Aplysia VAP333; synaptobrevin-associated protein of 33kDa; unnamed protein product; VAMP (vesicle-associated membrane protein)-associated protein A; VAMP (vesicle-associated membrane protein)-associated protein B and C; VAMP-A; VAMP-associated protein 33b; VAMP-associated protein A; VAMP-associated protein B; VAP-33; VAP-A; VAP33a; VAP33b; vesicle-associated membrane protein, associated protein a; vesicle-associated membrane protein, associated protein B and C
          View more View less
          Gene Aliases: 33kDa; D2Abb2e; VAMP-A; VAMP-B; Vamp33b; VAP-33; VAP-A; VAP-B; VAP33; VAP33b
          View more View less
          UniProt ID: (Mouse) Q9WV55, (Rat) Q9Z270, (Rat) Q9Z269
          View more View less
          Entrez Gene ID: (Mouse) 30960, (Rat) 58857, (Mouse) 56491, (Rat) 60431
          View more View less
          Function(s)
          protein binding microtubule binding protein domain specific binding FFAT motif binding identical protein binding protein homodimerization activity protein anchor macromolecular complex binding protein heterodimerization activity enzyme binding beta-tubulin binding
          Process(es)
          cellular calcium ion homeostasis ER to Golgi vesicle-mediated transport endoplasmic reticulum organization viral release from host cell cholesterol transport endoplasmic reticulum unfolded protein response IRE1-mediated unfolded protein response negative regulation by host of viral release from host cell negative regulation by host of viral genome replication positive regulation by host of viral genome replication modulation by host of viral RNA genome replication positive regulation of viral genome replication negative regulation by virus of viral protein levels in host cell COPII-coated vesicle budding endoplasmic reticulum membrane organization sphingomyelin biosynthetic process phospholipid transport sterol transport neuron projection development ceramide transport cholesterol homeostasis protein localization to endoplasmic reticulum protein folding in endoplasmic reticulum biological_process
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch