    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • MX1 Antibodies
          Proteintech
          MX1 Recombinant Rabbit Monoclonal Antibody (2L19)
          View all (25) MX1 antibodies
          Tech Support
          Cite MX1 Recombinant Rabbit Monoclonal Antibody (2L19)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (7)
          MX1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MX1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 7
          MX1 Antibody (82677-1-RR) in ICC/IF
          Immunofluorescent analysis of (-20°C Methanol) fixed A549 cells using MX1 antibody (82677-1-RR, Clone: 2L19) at dilution of 1:400 and CoraLite®488-Conjugated Goat Anti-Rabbit IgG (H+L) (SA00013-2). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MX1 Antibody in Immunocytochemistry (ICC/IF)
          MX1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MX1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MX1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MX1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MX1 Antibody in Western Blot (WB)
          MX1 Antibody in Surface plasmon resonance (SPR)
          MX1 Recombinant Rabbit Monoclonal Antibody (2L19)
          Product Details
          82677-1-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:500-1:2,000
          -
          Immunocytochemistry (ICC/IF)
          1:200-1:800
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          2L19
          Immunogen
          MX1 fusion protein Ag4692
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: LLEEGKATVP CLAEKLTSEL ITHICKSLPL LENQIKETHQ RITEELQKYG VDIPEDENEK MFFLIDKINA FNQDITALMQ GEETVGEEDI RLFTRLRHEF HKWSTIIENN FQEGHKILSR KIQKFENQYR GRELPGFVNY RTFETIVKQQ IKALEEPAVD MLHTVTDMVR LAFTDVSIKN FEEFFNLHRT AKSKIEDIRA EQEREGEKLI RLHFQMEQIV YCQDQVYRGA LQKVREKELE EEKKKKSWDF GAFQSSSATD SSMEEIFQHL MAYHQEASKR ISSHIPLIIQ FFMLQTYGQQ LQKAMLQLLQ DKDTYSWLLK ERSDTSDKRK FLKERLARLT QARRRLAQFP G
          Target Information
          This gene encodes a guanosine triphosphate (GTP)-metabolizing protein that participates in the cellular antiviral response. The encoded protein is induced by type I and type II interferons and antagonizes the replication process of several different RNA and DNA viruses. There is a related gene located adjacent to this gene on chromosome 21, and there are multiple pseudogenes located in a cluster on chromosome 4. Alternative splicing results in multiple transcript variants.
          Bioinformatics
          Protein Aliases: IFI-78K; Interferon-induced GTP-binding protein Mx1; Interferon-induced protein p78; interferon-inducible protein p78; Interferon-regulated resistance GTP-binding protein MxA; Myxoma resistance protein 1; myxovirus (influenza virus) resistance 1, interferon-inducible protein p78; Myxovirus resistance protein 1; p78 protein; unnamed protein product
          View more View less
          Gene Aliases: IFI-78K; IFI78; lncMX1-215; MX; MX1; MxA
          View more View less
          UniProt ID: (Human) P20591
          View more View less
          Entrez Gene ID: (Human) 4599
          View more View less
          Function(s)
          nucleotide binding GTPase activity protein binding GTP binding microtubule binding identical protein binding
          Process(es)
          immune system process apoptotic process defense response signal transduction response to virus synaptic vesicle budding from presynaptic endocytic zone membrane response to type I interferon negative regulation of viral genome replication innate immune response defense response to virus interleukin-27-mediated signaling pathway antiviral innate immune response
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline