Immunogen sequence: MSRRKPASGG LAASSSAPAR QAVLSRFFQS TGSLKSTSSS TGAADQVDPG AAAAAAAAAA TAPPAPPAPA FPPQLPPHIA TEIDRRKKRP LENDGPVKKK VKKVQQKEGG SDLGMSGNSE PKKCLRTRNV SKSLEKLKEF CCDSALPQSR VQTESLQERF AVLPKCTDFD DISLLHAKNA VSSEDSKRQI NQKDTTLFDL SQFGSSNTSH ENLQKTASKS ANKRSKSIYT PLELQYIEMK QQHKDAVLCV ECGYKYRFFG EDAEIAAREL NIYCHLDHNF MTASIPTHRL FVHVRRLVAK GYKVGVVKQT ETAALKAIGD NRSSLFSRKL TALYTKSTLI GEDVNPLIKL
Component of the post-replicative DNA mismatch repair system (MMR). Heterodimerizes with MSH2 to form MutS beta which binds to DNA mismatches thereby initiating DNA repair. When bound, the MutS beta heterodimer bends the DNA helix and shields approximately 20 base pairs. MutS beta recognizes large insertion-deletion loops (IDL) up to 13 nucleotides long. After mismatch binding, forms a ternary complex with the MutL alpha heterodimer, which is thought to be responsible for directing the downstream MMR events, including strand discrimination, excision, and resynthesis.
Protein Aliases: Divergent upstream protein; DNA mismatch repair protein Msh3; DUP; epididymis secretory sperm binding protein; hMSH3; Mismatch repair protein 1; MRP1; unnamed protein product
Gene Aliases: DUC1; DUG; DUP; FAP4; MRP1; MSH3
UniProt ID: (Human) P20585
Entrez Gene ID: (Human) 4437, (Rat) 499505