    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TAF15 Antibodies
          Proteintech
          TAF15 Recombinant Rabbit Monoclonal Antibody (3L13)
          View all (22) TAF15 antibodies
          Tech Support
          Cite TAF15 Recombinant Rabbit Monoclonal Antibody (3L13)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (8)
          TAF15 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TAF15 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 8
          TAF15 Antibody (82860-1-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HeLa cells using TAF15 antibody (82860-1-RR, Clone: 3L13) at dilution of 1:800 and Multi-rAb CoraLite ® Plus 594-Goat Anti-Rabbit Recombinant Secondary Antibody (H+L) (RGAR004).CoraLite® Plus 488-Phalloidin (green). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TAF15 Antibody in Immunocytochemistry (ICC/IF)
          TAF15 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TAF15 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TAF15 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TAF15 Antibody in Western Blot (WB)
          TAF15 Antibody in Western Blot (WB)
          TAF15 Antibody in Western Blot (WB)
          TAF15 Antibody in Surface plasmon resonance (SPR)
          TAF15 Recombinant Rabbit Monoclonal Antibody (3L13)
          Product Details
          82860-1-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:16,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:500-1:2,000
          -
          Immunocytochemistry (ICC/IF)
          1:400-1:1,600
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          3L13
          Immunogen
          TAF15 fusion protein Ag20828
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          0.8 mg/mL
          Amount
          80 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: GVSTDQVGEF FKQIGIIKTN KKTGKPMINL YTDKDTGKPK GEATVSFDDP PSAKAAIDWF DGKEFHGNII KVSFATRRPE FMRGGGSGGG RRGRGGYRGR GGFQGRGGDP KSGDWVCPNP SCGNMNFARR NSCNQCNEPR PEDSRPSGGD FRGRGYGGER GYRGRGGRGG DRGGYGGDRS GGGYGGDRSS GGGYSGDRSG GGYGGDRSGG GYGGDRGGGY GGDRGGGYGG DRGGGYGGDR GGYGGDRGGG YGGDRGGYGG DRGGYGGDRG GYGGDRGGYG GDRSRGGYGG DRGGGSGYGG DRSGGYGGDR SGGGYGGDRG GGYGGDRGGY GGKMGGRNDY RNDQRNRPY
          Target Information
          Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals.
          Bioinformatics
          Protein Aliases: 68 kDa TATA-binding protein-associated factor; hTAFII68; RBP56/CSMF fusion; RNA-binding protein 56; TAF(II)68; TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 68 kDa; TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 68kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68kD (RNA binding protein 56); TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68kD (RNA-binding protein 56); TATA box-binding protein-associated factor 2N (RNA-binding protein 56); TATA-bin; TATA-binding protein-associated factor 2N; TBP-associated factor 15; unnamed protein product
          View more View less
          Gene Aliases: 2610111C21Rik; 68kDa; Npl3; RBP56; TAF15; TAF2N; TAFII68
          View more View less
          UniProt ID: (Human) Q92804
          View more View less
          Entrez Gene ID: (Human) 8148, (Mouse) 70439, (Rat) 287571
          View more View less
          Function(s)
          nucleic acid binding DNA binding transcription cofactor activity RNA binding mRNA 3'-UTR binding protein binding zinc ion binding metal ion binding poly(A) RNA binding nucleotide binding molecular_function RNA metabolism protein
          Process(es)
          regulation of transcription, DNA-templated RNA splicing gene expression positive regulation of transcription, DNA-templated mRNA stabilization biological_process
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline